B3275
Monoclonal Anti-Rabbit Immunoglobulins–Biotin antibody produced in mouse
clone RG-16, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Monoclonal Anti-Rabbit Immunoglobulins (IgG, IgA, IgM)
About This Item
Recommended Products
biological source
mouse
conjugate
biotin conjugate
antibody form
purified immunoglobulin
antibody product type
secondary antibodies
clone
RG-16, monoclonal
form
buffered aqueous solution
species reactivity
rabbit
should not react with
human
pig, chicken, bovine, horse, human, guinea pig, turkey, rat, canine, goat, feline, sheep
technique(s)
direct ELISA: 1:60,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1,500
western blot: 1:200,000-1:400,000 using using an assay detecting actin in total cell extract of HeLa cells (5-10 μg per well)
western blot: 1:200,000
isotype
IgG1
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Looking for similar products? Visit Product Comparison Guide
General description
Specificity
Application
Immunohistochemistry (1 paper)
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service