Skip to Content
Merck
All Photos(1)

Key Documents

B2185

Sigma-Aldrich

Bisoprolol hemifumarate salt

≥98% (HPLC), solid

Synonym(s):

1-[4-[[2-(1-Methylethoxy)ethoxy]methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol hemifumarate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H31NO4 · .5C4H4O4
CAS Number:
Molecular Weight:
383.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: >20 mg/mL

originator

Teva

SMILES string

OC(=O)\C=C\C(O)=O.CC(C)NCC(O)COc1ccc(COCCOC(C)C)cc1.CC(C)NCC(O)COc2ccc(COCCOC(C)C)cc2

InChI

1S/2C18H31NO4.C4H4O4/c2*1-14(2)19-11-17(20)13-23-18-7-5-16(6-8-18)12-21-9-10-22-15(3)4;5-3(6)1-2-4(7)8/h2*5-8,14-15,17,19-20H,9-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

VMDFASMUILANOL-WXXKFALUSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

Application

Bisoprolol hemifumarate salt has been used as a β-adrenergic receptor (β-AR) antagonist in cardiac fibrosis mice model.

Biochem/physiol Actions

Bisoprolol hemifumarate is useful in oral formulations due to its high bioavailability. It also shows long elimination half-life.
Cardioselective β1-adrenoceptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Teva. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - STOT RE 2

Target Organs

Heart

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Tattersfield et al.
British journal of clinical pharmacology, 18(3), 343-347 (1984-09-01)
Bisoprolol is a new beta-adrenoceptor antagonist which has shown beta-adrenoceptor selectivity in studies in isolated tissues. Bronchial and cardiac beta-adrenoceptor blockade were assessed in eight normal subjects before and after oral ingestion of placebo, bisoprolol 20 and 40 mg, metoprolol
Cardiac beta-adrenergic receptor activation mediates distinct and cell type-dependent changes in the expression and distribution of connexin 43
Zhang Y, et al.
Journal of Cellular and Molecular Medicine (2020)
Pharmacokinetics and metabolism of bisoprolol enantiomers in humans
Horikiri Y, et al.
Journal of Pharmaceutical Sciences, 87(3), 289-294 (1998)
Viktor Nagy
Orvosi hetilap, 154(44), 1731-1734 (2013-10-29)
The prevalence of chronic heart failure in Hungary is 1.6% in the adult population, but it occurs in 15-20% of subjects over 80 years of age. The base of treatment of heart failure is the blockade of the neuro-hormonal system
Masaki Hashiyada et al.
Legal medicine (Tokyo, Japan), 15(2), 103-105 (2012-12-12)
An elderly person died of uncontrolled bradycardia in a hospital. The doctor had prescribed 1.35 mg of bisoprolol fumarate orally, but a nurse mistakenly gave the patient 10 mg of the drug 9 hours prior to her death. Bisoprolol was

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service