Skip to Content
Merck
All Photos(1)

Key Documents

B1502

Sigma-Aldrich

Brain Extract from bovine brain

Type I, Folch Fraction I

Synonym(s):

Brain Extract from Bovine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

biological source

bovine brain

type

Type I

form

powder

lipid type

phosphoglycerides

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Brain extract has lipids, especially polyunsaturated fatty acids (PUFAs) at high level. Extract from substantia niagra and the striatum has high amount of iron.

Application

Brain Extract from bovine brain has been used:
  • to prepare small unilamellar vesicles
  • in lipid binding assay
  • to aid the recovery of phosphoinositides and also to terminate the reaction in phosphatidylinositol 5-phosphate (PtdIns5P) and PtdIns4P assay
  • in liposome and protein self-aggregation assays

Biochem/physiol Actions

Brain lipids are involved in cellular signal transduction, membrane organization, trafficking protein modification, interactions and energy storage.

Packaging

Sealed ampule.

Components

Contains phosphatidylinositol phosphatidylserine,other brain lipids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of phosphoinositides in the zebrafish Danio rerio
Jones DR, et al.
Nature Protocols, 8(6), 1058-1058 (2013)
Lipid binding regulates synaptic targeting of PICK1, AMPA receptor trafficking, and synaptic plasticity
Jin W, et al.
The Journal of Neuroscience, 26, 2380-2390 (2006)
Structural and dynamic changes in human annexin VI induced by a phosphorylation-mimicking mutation, T356D
Freye-MC, et al.
Biochemistry, 42(3), 620-630 (2003)
Cholesterol and Fat Metabolism in Alzheimer?s Disease
Drug Discovery Approaches for the Treatment of Neurodegenerative Disorders, 161-193 (2017)
Structure and dynamics of helix-0 of the N-BAR domain in lipid micelles and bilayers
Low C, et al.
Biophysical Journal, 95(9), 4315-4323 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service