Skip to Content
Merck
All Photos(1)

Key Documents

B1125

Sigma-Aldrich

Bathocuproinedisulfonic acid disodium salt

Bathocuproinedisulfonic acid disodium salt

7.2-9.1% purity dry basis (ICP), powder

Synonym(s):

2,9-Dimethyl-4,7-diphenyl-1,10-phenantrolinedisulfonic acid disodium salt, Disodium bathocuproinedisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18N2Na2O6S2
CAS Number:
Molecular Weight:
564.54
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

Bathocuproinedisulfonic acid disodium salt, for spectrophotometric det. of Cu, Fe

Assay

7.2-9.1% dry basis (ICP)

Quality Level

form

powder

quality

for spectrophotometric det. of Cu, Fe

technique(s)

titration: 90% using HCLO4 (Dry Basis)

color

white to beige

solubility

H2O: 50 mg/mL

suitability

suitable for determination of copper

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1c(nc2c3nc(c(c(c3ccc2c1c5ccccc5)c4ccccc4)[S](=O)(=O)[O-])C)C

InChI

1S/C26H20N2O6S2.2Na/c1-15-25(35(29,30)31)21(17-9-5-3-6-10-17)19-13-14-20-22(18-11-7-4-8-12-18)26(36(32,33)34)16(2)28-24(20)23(19)27-15;;/h3-14H,1-2H3,(H,29,30,31)(H,32,33,34);;/q;2*+1/p-2

InChI key

RNGKZLRAVYPLJC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Bathocuproinedisulfonic acid is a strong copper chelator. It is used for the determination of copper and iron in biological samples.[1][2]

Application

  • Bathocuproinedisulfonic acid disodium salt has been used as a copper chelator for specific assay in Arabidopsis seedlings.[3]
  • It has been used as an electron chain-associated free radical production inhibitor for superoxide dismutase assay in cells.[4]
  • It has been added to the culture media of trypanosomes as a copper chelator for the stabilization of cysteine.[5]

Other Notes

Mixture of isomers

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minglin Lang et al.
Neurobiology of aging, 34(11), 2604-2612 (2013-07-06)
Disruption of copper homeostasis has been implicated in Alzheimer's disease (AD) during the last 2 decades; however, whether copper is a friend or a foe is controversial. Within a genetically tractable Drosophila AD model, we manipulated the expression of human
Jennifer Lowe et al.
The Journal of biological chemistry, 279(25), 25986-25994 (2004-04-14)
Ccc2p is homologous to the human Menkes and Wilson copper ATPases and is herein studied as a model for human copper transport. Most studies to date have sought to understand how mutations in the human Menkes or Wilson genes impair
Shinya Ogawa et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 909, 34-36 (2012-11-17)
HPLC eluent systems employing acetonitrile and methanol were evaluated for the quantitation of glutathione (GSH) and phytochelatin (PC(n)), a family of peptides implicated in heavy-metal detoxification in higher plants. The detection system is based on the dequenching of copper(I)-bathocuproine disulfonate
Measurement of superoxide dismutase, catalase and glutathione peroxidase in cultured cells and tissue.
Weydert CJ and Cullen JJ
Nature Protocols, 5, 51-66 (2010)
Shinya Ogawa et al.
Analytical chemistry, 81(21), 9199-9200 (2009-09-30)
Reoptimization of analytical conditions was performed for a high-performance liquid chromatographic (HPLC) detection system for Cu(I) chelators based on the dequenching of Cu(I)-bathocuproine disulfonate complexes that occurs in the presence of Cu(I) chelators. The revision corrects for emission and excitation

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service