Skip to Content
Merck
All Photos(1)

Key Documents

B112

Sigma-Aldrich

(±)-Bay K8644

calcium channel agonist

Synonym(s):

1,4-Dihydro-2,6-dimethyl-5-nitro-4-(2-[trifluoromethyl]phenyl)pyridine-3-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15F3N2O4
CAS Number:
Molecular Weight:
356.30
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥97.5% (HPLC)

form

powder

color

yellow to dark yellow

mp

168-170  °C

solubility

DMSO: 20 mg/mL
methanol and ethanol: 63 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

CC1=C(C(OC)=O)C(C2=CC=CC=C2C(F)(F)F)C([N+]([O-])=O)=C(C)N1

InChI

1S/C16H15F3N2O4/c1-8-12(15(22)25-3)13(14(21(23)24)9(2)20-8)10-6-4-5-7-11(10)16(17,18)19/h4-7,13,20H,1-3H3

InChI key

ZFLWDHHVRRZMEI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Bay K8644 has been used as an agonist for voltage sensitive calcium channels (VSCCs) in interneurons,[1] L-type calcium channel (LTCC) in human embryonic kidney 293 cells[2] and viscerofugal neurons.[3]

Biochem/physiol Actions

(±)-Bay K8644 is a 1,4-dihydropyridine (DHP) derivative and an agonist for L-type Ca2+ channels.[4] However, it′s enantiomers (-)-BAY k8644 and (+)-BAY k8644 have opposing functionality with respect to Ca2+ channels as high-potency agonist and low-potency antagonist, respectively.[5] (±)-Bay K8644 increases influx of Ca2+ specifically at voltage-gated calcium channels. It constricts blood vessels and heart contraction.[6]
Increases influx of Ca2+ specifically at voltage-gated calcium channels.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CaV1.2 channelopathic mutations evoke diverse pathophysiological mechanisms.
Bamgboye, et al.
The Journal of General Physiology, 154 (2023)
Opposing actions of Bay K 8644 enantiomers on calcium current, prolactin secretion, and synthesis in pituitary cells
Enyeart JJ, et al.
Molecular Endocrinology, 4(5), 727-735 (1990)
Spectrum of Cav1. 4 dysfunction in congenital stationary night blindness type 2
Burtscher V, et al.
Biochimica et Biophysica Acta, 1838(8), 2053-2065 (2014)
Diego Santos de Souza et al.
European journal of pharmacology, 859, 172488-172488 (2019-06-25)
Cardiac arrhythmias are among the most important pathologies that lead to sudden death. The discovery of new therapeutic options against arrhythmias with low adverse effects is of paramount importance. Farnesol is found in essential oils with antioxidant, anti-inflammatory and cardioprotective
The calcium channel agonist BAY k 8644 reduces ethanol intake and preference in alcohol-preferring AA rats
de Beun R, et al.
Psychopharmacology, 127(1-2), 302-310 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service