AXYST200SSX
Screw cap tubes without caps, conical bottom, self-standing
size 2.0 mL, amber, pkg of 8x500caps/cs
Synonym(s):
Screw cap tubes without caps, conical bottom, self-standing
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
Recommended Products
material
conical bottom tube (standing)
polypropylene
sterility
non-sterile
feature
graduations: no
manufacturer/tradename
Axygen Inc ST-200-SS-X
Corning
packaging
pkg of 8x500caps/cs
size
2.0 mL
color
amber
Looking for similar products? Visit Product Comparison Guide
General description
Product is only available in China.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service