Skip to Content
Merck
All Photos(1)

Key Documents

AXYRFL222C

Pipette tips refill rack

for universal fit tips, graduated, capacity 200 μL, pkg of 5x960tips/cs, clear

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.02
volume:
200 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc RFL-222-C
Corning

manufacturer/tradename

Axygen Inc RFL-222-C
Corning

packaging

pkg of 5x960tips/cs

capacity

200 μL

volume

200 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Browse all pipette tips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service