Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR0104C

PCR strip tubes

with strip caps, 4 per strip, size 0.1 mL, clear, pkg of 10x250tubes/cs

Synonym(s):

96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.11

material

cap (plugseal)
polypropylene

sterility

non-sterile

manufacturer/tradename

Axygen Inc PCR-0104-C
Corning

packaging

pkg of 10x250tubes/cs

size

0.1 mL

color

clear
clear

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Made from 99.9% pure virgin polypropylene
  • RNase-/DNase-free and nonpyrogenic
  • Compatible with the Corbett rotor gene system

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service