Skip to Content
Merck
All Photos(3)

Key Documents

AV53716

Sigma-Aldrich

Anti-ISYNA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Myo-inositol 1-phosphate synthase A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ISYNA1(51477)

Immunogen

Synthetic peptide directed towards the N terminal region of human ISYNA1

Application

Anti-ISYNA1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ISYNA1 (inositol-3-phosphate synthase 1) gene encodes a protein that belongs to myo-inositol 1-phosphate synthase family and is predominantly expressed in testis, ovary, heart, placenta and pancreas. It is a crucial enzyme in myo-inositol biosynthesis pathway that plays a pivotal role in catalyzing the rate-limiting conversion of glucose 6-phosphate to myoinositol 1-phosphate.

Sequence

Synthetic peptide located within the following region: LQEQLWPHMEALRPRPSVYIPEFIAANQSARADNLIPGSRAQQLEQIRRD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guimin Guan et al.
Archives of biochemistry and biophysics, 417(2), 251-259 (2003-08-28)
myo-Inositol 1-phosphate synthase (EC 5.5.1.4) (IPS) is a key enzyme in myo-inositol biosynthesis pathway. This study describes the molecular cloning of the full length human myo-inositol 1-phosphate synthase (hIPS) cDNA, tissue distribution of its mRNA and characterizes its gene expression
Shulin Ju et al.
The Journal of biological chemistry, 279(21), 21759-21765 (2004-03-17)
We have cloned, sequenced, and expressed a human cDNA encoding 1-d-myo-inositol-3-phosphate (MIP) synthase (hINO1). The encoded 62-kDa human enzyme converted d-glucose 6-phosphate to 1-d-myo-inositol 3-phosphate, the rate-limiting step for de novo inositol biosynthesis. Activity of the recombinant human MIP synthase
A Azimi et al.
British journal of cancer, 110(10), 2489-2495 (2014-04-12)
Disseminated cutaneous malignant melanoma (CMM) is commonly unresponsive to standard chemotherapies, and there are as yet no predictive markers of therapy response. In the present study we collected fresh-frozen pretreatment lymph-node metastasis samples (n=14) from melanoma patients with differential response

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service