Skip to Content
Merck
All Photos(2)

Key Documents

AV44743

Sigma-Aldrich

Anti-TGFBR2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-HNPCC6, Anti-MFS2, Anti-RIIC, Anti-Tbeta, Anti-Transforming growth factor, β receptor II (70/80 kDa)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

62 kDa

species reactivity

rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGFBR2(7048)

General description

Transforming growth factor, β receptor II (70/80 kDa) (TGFBR2, HNPCC6, MFS2, RIIC) is a transmembrane ser/thr protein kinase family receptor for TGF-β (TGFB). TGFBR2 mediate TGF-β cell signaling to regulate/inhibit cell proliferation. Defective TGFBR2 is associated with Marfan syndrome.

Specificity

Anti-TGFBR2 polyclonal antibody reacts with bovine, rabbit, human, mouse, rat, and pig transforming growth factor, β receptor II proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human TGFBR2

Application

Anti-TGFBR2 polyclonal antibody is used to tag transforming growth factor, β receptor II for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of transforming growth factor, β receptor II in TGF-β signaling and regulation of cell proliferation.

Biochem/physiol Actions

TGFBR2 is a member of the Ser/Thr protein kinase family and the TGFB receptor subfamily. The protein is a transmembrane protein that has a protein kinase domain, forms a heterodimeric complex with another receptor protein, and binds TGF-beta. This receptor/ligand complex phosphorylates proteins, which then enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation. Mutations in its gene have been associated with Marfan Syndrome, Loeys-Deitz Aortic Aneurysm Syndrome, and the development of various types of tumors.This gene is a member of the Ser/Thr protein kinase family and the TGFB receptor subfamily. It encodes a transmembrane protein that has a protein kinase domain, forms a heterodimeric complex with another receptor protein and binds TGF-beta. This receptor/ligand complex phosphorylates proteins which then enter the nucleus and regulate the transcription of a subset of genes related to cell proliferation. Mutations in this gene have been associated with Marfan Syndrome, Loeys-Deitz Aortic Aneurysm Syndrome, and the development of various types of tumors. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

Sequence

Synthetic peptide located within the following region: MGRGLLRGLWPLHIVLWTRIASTIPPHVQKSDVEMEAQKDEIICPSCNRT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Yang et al.
Cell death & disease, 10(8), 558-558 (2019-07-25)
Natriuretic peptide type C (NPPC) secreted by mural granulosa cells (MGCs) maintains oocyte meiotic arrest via the activation of guanylyl cyclase-linked natriuretic peptide receptor 2 (NPR2). Here, we investigated the effect of transforming growth factor (TGF)-β on NPPC expression in
Luke D Halder et al.
Nature communications, 11(1), 2331-2331 (2020-05-13)
Extracellular vesicles have an important function in cellular communication. Here, we show that human and mouse monocytes release TGF-β1-transporting vesicles in response to the pathogenic fungus Candida albicans. Soluble β-glucan from C. albicans binds to complement receptor 3 (CR3, also
Peng Li et al.
Stem cell research & therapy, 10(1), 144-144 (2019-05-23)
Non-small cell lung cancer (NSCLC) is the second most prevalent cause of cancer-related fatality. Long non-coding RNAs (lncRNAs) have been observed to exercise functions in NSCLC. Here, the current study aimed to explore the potential mechanism of lncRNA MBNL1-AS1 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service