Skip to Content
Merck
All Photos(5)

Key Documents

AV42623

Sigma-Aldrich

Anti-CHAC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ChaC, cation transport regulator homolog 1 (E. coli), Anti-MGC4504

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
pig, rabbit, human, rat, mouse, dog, bovine, horse
citations:
4
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

pig, rabbit, human, rat, mouse, dog, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHAC1(79094)

General description

CHAC1/MGC4504 (cation transport regulator-like protein 1), a novel gene regulated by the atherosclerotic lesion component ox-PAPC (oxidized 1-palmitoyl-2-arachidonyl-sn-3-glycero-phosphorylcholine), is a proapoptotic component of the ATF4-ATF3-CHOP cascade mediating the unfolded protein response pathway within cells.

Specificity

Anti-CHAC1 polyclonal antibody reacts with human, canine, bovine, rat, mouse, mouse, rat, and human cation transport regulator-like protein 1 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human CHAC1

Application

Anti-CHAC1 polyclonal antibody is used to tag cation transport regulator-like protein 1 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of CHAC1 in the regulation of apoptosis via the unfolded protein response pathway.

Biochem/physiol Actions

CHAC1 belongs to the chaC family. The exact function of CHAC1 remains unknown.

Sequence

Synthetic peptide located within the following region: TPQNPGYLGPAPEEAIATQILACRGFSGHNLEYLLRLADFMQLCGPQAQD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nam E Joo et al.
Cancer medicine, 1(3), 295-305 (2013-01-24)
Nisin, a bacteriocin and commonly used food preservative, may serve as a novel potential therapeutic for treating head and neck squamous cell carcinoma (HNSCC), as it induces preferential apoptosis, cell cycle arrest, and reduces cell proliferation in HNSCC cells, compared
Léa Perra et al.
Frontiers in immunology, 9, 2823-2823 (2018-12-18)
In cystic fibrosis (CF), Pseudomonas aeruginosa (Pa) colonizes the lungs, leading to chronic inflammation of the bronchial epithelium. ChaC glutathione-specific γ-glutamylcyclotransferase 1 (CHAC1) mRNA is differentially expressed in primary human airway epithelial cells from bronchi (hAECBs) from patients with CF
Amandeep Kaur et al.
The Journal of biological chemistry, 292(2), 638-651 (2016-12-04)
Glutathione degradation plays an important role in glutathione and redox homeostasis, and thus it is imperative to understand the enzymes and the mechanisms involved in glutathione degradation in detail. We describe here ChaC2, a member of the ChaC family of
Willian Meira et al.
Cancers, 13(6) (2021-04-04)
In our previous study, we showed that a cystine transporter (xCT) plays a pivotal role in ferroptosis of pancreatic ductal adenocarcinoma (PDAC) cells in vitro. However, in vivo xCTKO cells grew normally indicating that a mechanism exists to drastically suppress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service