Skip to Content
Merck
All Photos(1)

Key Documents

AV39663

Sigma-Aldrich

Anti-MXD3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MAD3, Anti-MAX dimerization protein 3, Anti-MGC2383

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

23 kDa

species reactivity

horse, mouse, guinea pig, bovine, rat, human, rabbit, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MXD3(83463)

General description

MXD3 is a basic, helix-loop-helix transcription factor that belongs to the MAD family of proteins. This protein is involved in cerebellar development and GNP proliferation. Studies have reported that MXD3 is also required for the proliferation of DAOY medulloblastoma cells.
Rabbit Anti-MXD3 antibody binds to chicken, human, mouse, rat, bovine, zebrafish, and canine MXD3.

Immunogen

Synthetic peptide directed towards the N terminal region of human MXD3

Application

Rabbit Anti-MXD3 antibody is suitable for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

MXD3 contains 1 basic helix-loop-helix (bHLH) domain. It is a transcriptional repressor and binds with MAX to form a sequence-specific DNA-binding protein complex which recognizes the core sequence 5′-CAC[GA]TG-3′. Antagonizes MYC transcriptional activity by competing for MAX and suppresses MYC dependent cell transformation.

Sequence

Synthetic peptide located within the following region: MEPLASNIQVLLQAAEFLERREREAEHGYASLCPHRSPGPIHRRKKRPPQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gustavo A Barisone et al.
PloS one, 7(7), e38508-e38508 (2012-07-19)
A subset of medulloblastomas, the most common brain tumor in children, is hypothesized to originate from granule neuron precursors (GNPs) in which the sonic hedgehog (SHH) pathway is over-activated. MXD3, a basic helix-look-helix zipper transcription factor of the MAD family

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service