Skip to Content
Merck
All Photos(1)

Key Documents

AV31955

Sigma-Aldrich

Anti-HOXA13 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HOX1, Anti-HOX1J, Anti-Homeobox A13

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HOXA13(3209)

General description

HOXA13 is a homeobox transcription factor involved in embryonic development. This transcription factor is also involved in digit patterning in mice. Mutations in HOXA13 have been linked to hand-foot-genital syndrome.
Rabbit Anti-HOXA13 antibody recognizes human HOXA13.

Immunogen

Synthetic peptide directed towards the N terminal region of human HOXA13

Application

Rabbit Anti-HOXA13 antibody can be used for western blot applications at 2.5μg/ml.

Biochem/physiol Actions

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Expansion of a polyalanine tract in the encoded protein can cause hand-foot-uterus syndrome, also known as hand-foot-genital syndrome. In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Expansion of a polyalanine tract in the encoded protein can cause hand-foot-uterus syndrome, also known as hand-foot-genital syndrome.

Sequence

Synthetic peptide located within the following region: AAAAANQCRNLMAHPAPLAPGAASAYSSAPGEAPPSAAAAAAAAAAAAAA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D P Mortlock et al.
Nature genetics, 15(2), 179-180 (1997-02-01)
There are several human syndromes which involve defects of the limbs and the Müllerian ducts or its derivatives. The hand-foot-genital (HFG) syndrome is an autosomal dominant, fully penetrant disorder that was originally described by Stern et al. Additional reports describing
Keng Yih Chew et al.
BMC developmental biology, 12, 2-2 (2012-01-13)
Kangaroos and wallabies have specialised limbs that allow for their hopping mode of locomotion. The hindlimbs differentiate much later in development but become much larger than the forelimbs. The hindlimb autopod has only four digits, the fourth of which is
Ting-Ting Pan et al.
The Tohoku journal of experimental medicine, 234(3), 209-219 (2014-10-25)
HOXA13 is a member of homeobox genes that encode transcription factors regulating embryonic development and cell fate. Abnormal HOXA13 expression was reported in hepatocellular carcinoma (HCC), but its correlation with tumor angiogenesis and prognosis still remain unclear. This study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service