Skip to Content
Merck
All Photos(4)

Documents

A9883

Sigma-Aldrich

ADA

≥98% (titration)

Synonym(s):

N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCOCH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
190.15
Beilstein:
1787181
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (titration)

form

powder

pH

1.5-3.0

useful pH range

6.0-7.2

pKa (25 °C)

6.6

mp

219 °C (dec.) (lit.)

solubility

1 M NaOH: 160 mg/mL, clear, colorless

application(s)

diagnostic assay manufacturing

storage temp.

room temp

SMILES string

NC(=O)CN(CC(O)=O)CC(O)=O

InChI

1S/C6H10N2O5/c7-4(9)1-8(2-5(10)11)3-6(12)13/h1-3H2,(H2,7,9)(H,10,11)(H,12,13)

InChI key

QZTKDVCDBIDYMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ADA is a zwitterionic buffer used in biochemistry and molecular biology. It is one of the Good buffers developed in the 1960′s to provide buffers in the pH range of 6.15 - 8.35 for wide applicability to biochemical studies. The pioneering publication by Good and co-workers describes the synthesis of ADA and its physical properties. The useful range of ADA buffer in aqueous solution is 6.0 - 7.2.

Application

Biological buffer component with sodium hydroxide and sodium chloride (pH 5.67-7.57, useful pH range 6.4-7.4). Used to prepare immobilized pH gradients.

Preparation Note

This product is soluble in 1 M NaOH (160mg/mL), yielding a clear, colorless solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrogen ion buffers for biological research
Good NE, et al.
Biochemistry, 5( (2), 467-477 (1966)
Harold Bays et al.
The Journal of clinical endocrinology and metabolism, 100(8), 3140-3148 (2015-06-02)
Despite current standard of care, many patients at high risk of cardiovascular disease (CVD) still have elevated low-density lipoprotein cholesterol (LDL-C) levels. Alirocumab is a fully human monoclonal antibody inhibitor of proprotein convertase subtilisin/kexin type 9. The objective of the
Johannan F Brandse et al.
Journal of Crohn's & colitis, 9(11), 973-981 (2015-06-28)
High serum concentrations of infliximab [IFX] and adalimumab [ADA] may be associated with adverse effects in patients with inflammatory bowel disease [IBD]. We aimed to investigate whether high anti-tumour necrosis factor [TNF] trough levels [TLs] were associated with toxicity and
Gulen Yesilbag Tonga et al.
Nature chemistry, 7(7), 597-603 (2015-06-24)
Bioorthogonal catalysis broadens the functional possibilities of intracellular chemistry. Effective delivery and regulation of synthetic catalytic systems in cells are challenging due to the complex intracellular environment and catalyst instability. Here, we report the fabrication of protein-sized bioorthogonal nanozymes through
Christopher J Smiley et al.
Journal of the American Dental Association (1939), 146(7), 508-524 (2015-06-27)
Conduct a systematic review and meta-analysis on nonsurgical treatment of patients with chronic periodontitis by means of scaling and root planing (SRP) with or without adjuncts. A panel of experts convened by the American Dental Association Council on Scientific Affairs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service