Skip to Content
Merck
All Photos(2)

Key Documents

A9669

Sigma-Aldrich

Anti-Human IgA (α-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgA (α-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:35,000
dot blot: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin A (IgA) is the most abundant antibody isotype produced in humans. IgAs are heterogeneous and exist as two IgA subtypes. It has the ability to form multimers by interacting with the joining (J) chain.It is present in large quantities in blood. IgA antibodies protect mucosal surfaces against invading pathogens.
Specificity of the Alkaline Phosphatase Conjugated Anti-Human IgA is determined by Enzyme Linked Immunosorbent Assay (ELISA). Identity and purity of the antibody is established by immunoelectrophoresis (IEP) prior to conjugation. Electrophoresis of the antibody preparation followed by diffusion versus anti-goat IgG and anti-goat whole serum results in single arcs of precipitation.

Specificity

The conjugate is specific for human IgG. No cross-reaction with Mouse and Rat IgG.

Immunogen

Purified human IgA

Application

Anti-Human IgA (α-chain specific) Alkaline Phosphatase antibody produced in goat has been used in quantitative sandwich enzyme-linked immunosorbent assay (ELISA) and immunoassays.
Human milk samples were analyzed for the presence of IgA antibodies specific to certain antigens of H. pylori by western blot using alkaline-phosphate-conjugated goat anti-human IgA antibody.
Milk samples from 10 mothers were probed for bound milk IgA by western blot using alkaline-phosphatase goat anti-human IgA antibody.Elisa Assays were performed using alkaline phosphatase conjugated gaot anti-human IgA chain specific antibody (1:1000) to detect IgA immunoglobulins in serum and seminal plasma samples from infertile men.
The product may also be used for dot blot (1:30,000) and immunohistochemistry (at 1:100 dilutions, using formalin-fixed, paraffin-embedded sections).

Biochem/physiol Actions

IgA is a secretory glycoprotein antibody that regulates the physiological balance between commensal intestinal bacteria and the defenses of the host immune system. It is the highest generated antibody isotype. IgAs are mainly secreted in the intestinal mucosa and prevent the commensal microorganisms from invading the intestines
Immunoglobulin A (IgA) antibody is a secretory antibody present abundantly in mucous linings of gastrointestinal, respiratory and genitourinary tracts, tears and saliva. Thus, secretory IgA contributes to the humoral defense mechanism against the pathogens on mucosal surfaces.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide

Storage and Stability

Store at 0-5°C. Do Not Freeze.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian E Thomas et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 39(8), 1155-1160 (2004-10-16)
Immunoglobulin A (IgA) in maternal milk may protect Gambian infants from early Helicobacter pylori colonization. This study sought evidence that this protection could be due to specific IgA antibodies. Sixty-five infants were screened from 12 weeks of age with [13C]-urea
Luka Mesin et al.
Journal of immunology (Baltimore, Md. : 1950), 187(6), 2867-2874 (2011-08-16)
To understand the biology of Ab-secreting cells in the human small intestine, we examined Ab production of intestinal biopsies kept in culture. We found sustained IgA and IgM secretion as well as viable IgA- or IgM-secreting cells after >4 wk
Mucosal immunity and vaccines
Holmgren J and Czerkinsky C
Nature Medicine, 11(4s), S45-S45 (2005)
Elena Mitsi et al.
The Journal of clinical investigation, 132(7) (2022-02-10)
BackgroundAlthough recent epidemiological data suggest that pneumococci may contribute to the risk of SARS-CoV-2 disease, cases of coinfection with Streptococcus pneumoniae in patients with coronavirus disease 2019 (COVID-19) during hospitalization have been reported infrequently. This apparent contradiction may be explained
Esha Oommen et al.
Clinical and experimental rheumatology, 35 Suppl 103(1), 98-101 (2017-03-11)
To determine the prevalence of anti-myeloperoxidase (MPO) antibodies of IgA (IgA anti-MPO) isotype in patients with eosinophilic granulomatosis with polyangiitis (EGPA), and the association of the IgA antibodies with IgG anti-MPO and with disease activity. Serum samples from patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service