Skip to Content
Merck
All Photos(2)

Key Documents

A8667

Sigma-Aldrich

Anti-Human IgG (whole molecule)−Peroxidase antibody produced in goat

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Goat Anti-Human IgG (whole molecule) HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct ELISA: 1:30,000
dot blot: 1:80,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders.
Goat Anti-Human IgG (whole molecule)-Peroxidase antibody binds to human IgG.

Immunogen

human IgG

Application

Anti-Human IgG (whole molecule)-Peroxidase antibody produced in goat has also been used as a conjugate for gold nanoparticles (AuNPs) in electrochemical detection.
Goat Anti-Human IgG (whole molecule)-Peroxidase antibody has been used for ELISA, dot blot and immunohistochemistry.
Specificities of anti-human HIV-1 envelope antibodies was determined by ELISA using HRP-conjugated goat anti-human IgG at a dilution of 1:1000.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marzieh Ashrafmansouri et al.
Iranian journal of parasitology, 10(4), 599-604 (2016-01-27)
Cutaneous leishmaniasis (CL) is a parasitic disease with a relatively wide distribution in different areas of the world, including Iran. The parasite is mainly diagnosed microscopically, but serological approaches might be useful for diagnosis as well. This study aimed to
Elin S Gray et al.
Journal of virology, 83(17), 8925-8937 (2009-06-26)
Defining the specificities of the anti-human immunodeficiency virus type 1 (HIV-1) envelope antibodies able to mediate broad heterologous neutralization will assist in identifying targets for an HIV-1 vaccine. We screened 70 plasmas from chronically HIV-1-infected individuals for neutralization breadth. Of
Leiguang Ye et al.
FEBS open bio, 5, 809-812 (2015-11-14)
An in-house enzyme-linked immunosorbent assay (ELISA) was developed in this study to detect circulating IgG antibodies to peptide antigens derived from baculoviral IAP repeat-containing protein 5 isoform 2 (BIRC5) and myc proto-oncogene protein (MYC) in non-small cell lung cancer (NSCLC).
L D Curtis et al.
Journal of thrombosis and haemostasis : JTH, 3(6), 1180-1186 (2005-05-17)
BB-10153 is an engineered variant of human plasminogen that is activated to plasmin by thrombin. Thrombus-selective induction of reperfusion and prevention of reocclusion have been demonstrated following bolus administration in animal models of thrombosis. The objective of the study was
Hui Wang et al.
Virology journal, 8, 539-539 (2011-12-16)
Kaposi sarcoma (KS) is a complex multifocal neoplasm and is the major cause of death for about 50% of acquired immunodeficiency syndrome (AIDS) patients. Kaposi's sarcoma-associated herpesvirus (KSHV) is an oncogenic virus with a causal role in the development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service