Skip to Content
Merck
All Photos(2)

Key Documents

A7310

Sigma-Aldrich

Anti-Water Channel Aquaporin 2 antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-Aquaporin 2, Aquaporin 2 Detection, Rabbit Anti-Aquaporin 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot (chemiluminescent): 1:200-1:1000

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AQP2(359)
mouse ... Aqp2(11827)
rat ... Aqp2(25386)

General description

Aquaporin 2 (AQP2) exists as a tetramer with C terminus showing multiple conformations and binds two cadmium ions. AQP2 gene is mapped to human chromosome 12q13.12. It is associated with intracellular vesicles.
Aquaporin 2 (AQP2) is a water channel protein that regulates water balance in renal connecting tubules and collecting ducts. AQP2 is activated by anti-diuretic hormone and subsequently facilitates water inflow in renal cells. This protein also interacts with integrin to modulate epithelial cell morphogenesis and cell movement . Anti-Water Channel Aquaporin 2 antibody recognizes aquaporin 2 from rat by immunoblotting. The antibody also reacts with mouse and human AQP2.

Immunogen

synthetic peptide corresponding to amino acids 254-271 of rat or mouse aquaporin 2 (with additional N-terminal lysine and tyrosine) conjugated to KLH with glutaraldehyde. The epitope is identical in rat and mouse and highly conserved in human.

Application

Anti-Water Channel Aquaporin 2 antibody is also suitable for use in chemiluminescent western blot (1:200-1:1000), and immunoblot.
Anti-Water Channel Aquaporin 2 antibody produced in rabbit has been used in:
  • immunostaining
  • immunohistochemistry
  • immunofluorescence
  • immunoblotting

Immunohistochemistry was performed on 10% formalin-fixed parraffin embedded rat kidney sections using rabbit anti-AQP2 at a concentration of 5μg/ml. Use of anti-AQP2 allowed visualization of the medullary thick ascending limb and collecting ducts.

Biochem/physiol Actions

Aquaporin (AQP2) N-terminal region plays a key role in trafficking. The C-terminal phosphorylation is crucial for the water permeability function of the apical membrane. Also, ubiquitination at lysine 270 is needed for AQP2 sorting and degradation in lysosomes or elimination through exosomes. Mutation in AQP2 gene is implicated in congenital nephrogenic diabetes insipidus (NDI), a water balance disorder. The functional dysregulation of AQP2 is also correlated to preeclampsia, congestive heart failure and liver cirrhosis.

Physical form

Lyophilized from a solution containing phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide.

Preparation Note

Isolated on immobilized Protein A

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Rieger et al.
PloS one, 11(7), e0158977-e0158977 (2016-07-16)
During nephrogenesis, POU domain class 3 transcription factor 3 (POU3F3 aka BRN1) is critically involved in development of distinct nephron segments, including the thick ascending limb of the loop of Henle (TAL). Deficiency of POU3F3 in knock-out mice leads to
Kuniaki Takata et al.
Histochemistry and cell biology, 130(2), 197-209 (2008-06-21)
Aquaporins (AQPs) are membrane proteins serving in the transfer of water and small solutes across cellular membranes. AQPs play a variety of roles in the body such as urine formation, prevention from dehydration in covering epithelia, water handling in the
Missense mutation of POU domain class 3 transcription factor 3 in Pou3f3L423P mice causes reduced nephron number and impaired development of the thick ascending limb of the loop of Henle
Rieger A, et al.
Testing, 11(7) (2016)
Hydrochlorothiazide ameliorates polyuria caused by tolvaptan treatment of polycystic kidney disease in PCK rats
Wang A, et al.
Clinical and Experimental Nephrology, 23(4), 455-464 (2019)
Surface glycosylation profiles of urine extracellular vesicles
Gerlach JQ, et al.
PLoS ONE, 8(9) (2013)

Questions

  1. Does the Product No. A7310, Anti-Water Channel Aquaporin 2 antibody, cross-react with the AQP1 protein?

    1 answer
    1. The cross-reaction of this antibody with AQP1 has not been tested. The immunogen sequence used to make Product No. A7310, Anti-Water Channel Aquaporin 2, is the rat AQP2 sequence: RQSVELHSPQSLPRGSKA. There is no homology between this sequence and rat AQP1, so cross-reaction would not be expected.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service