Skip to Content
Merck
All Photos(1)

Key Documents

A7106

Sigma-Aldrich

Azelnidipine

≥98% (HPLC), powder

Synonym(s):

2-Amino-1,4-dihydro-6-methyl-4-(3 nitrophenyl)-3,5-pyridinedicarboxylic acid 3-[1-(diphenylmethyl)-3-azetidinyl] 5-(1-methylethyl) ester, CS-905, RS-9054

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N4O6
CAS Number:
Molecular Weight:
582.65
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >10 mg/mL

originator

Daiichi-Sankyo

storage temp.

room temp

SMILES string

CC(C)OC(=O)C1=C(C)NC(N)=C(C1c2cccc(c2)[N+]([O-])=O)C(=O)OC3CN(C3)C(c4ccccc4)c5ccccc5

InChI

1S/C33H34N4O6/c1-20(2)42-32(38)27-21(3)35-31(34)29(28(27)24-15-10-16-25(17-24)37(40)41)33(39)43-26-18-36(19-26)30(22-11-6-4-7-12-22)23-13-8-5-9-14-23/h4-17,20,26,28,30,35H,18-19,34H2,1-3H3

InChI key

ZKFQEACEUNWPMT-UHFFFAOYSA-N

Application

Azelnidipine is a novel dihydropyridine derivative, a L-type calcium channel blocker, and an antihypertensive. Acute administration of azelnidipine prevents a sudden drop of cardiac function after acute stress. Azelnidipine may have a protective role in inflammation associated with atherosclerosis.

Biochem/physiol Actions

Azelnidipine, a novel dihydropyridine derivative, is a L-type calcium channel blocker and antihypertensive. Unlike other L-type calcium channel blockers, azelnidipine causes minimal stimulation of the sympathetic nervous system despite its significant depressor effect. Azelnidipine may have a protective role in inflammation in atherosclerosis.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Daiichi-Sankyo. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Matsui et al.
Hypertension (Dallas, Tex. : 1979), 59(6), 1132-1138 (2012-05-02)
Day-by-day home blood pressure (BP) variability (BPV) was reported to be associated with increased cardiovascular risk. We aimed to test the hypothesis that the angiotensin II receptor blocker/calcium-channel blocker combination decreases day-by-day BPV more than the angiotensin II receptor blocker/diuretic
Yoshio Matsui et al.
American journal of hypertension, 25(8), 862-868 (2012-06-01)
We aimed to investigate the association of the change in the ambulatory arterial stiffness index (AASI) with that in carotid-femoral pulse wave velocity (cfPWV) during treatment with antihypertensive medication. We enrolled 207 hypertensive patients treated with olmesartan monotherapy for 12
Takeshi Takami et al.
Vascular health and risk management, 7, 383-390 (2011-07-29)
The aim of this study was to compare the effects of olmesartan combined with either azelnidipine or amlodipine on central blood pressure (CBP) and left ventricular mass index (LVMI) in hypertensive patients. Patients with brachial systolic BP ≥ 140 mmHg
Takehisa Shimizu et al.
International heart journal, 53(5), 331-335 (2012-10-06)
Vascular calcification is an active and regulated process that is similar to bone formation. While calcium channel blockers (CCBs) have been shown to improve outcomes in atherosclerotic vascular disease, it remains unknown whether CCBs have an effect on the process
Ikuo Saito et al.
Journal of nephrology, 25(5), 699-708 (2011-10-25)
In the present investigation we extracted data on hypertensive patients with chronic kidney disease (CKD) who were enrolled in 3 studies - 2 studies of the angiotensin receptor blocker (ARB) olmesartan medoxomil (OLM), lasting 12 weeks and 2 years, respectively

Articles

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

We offers many products related to calcium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service