Skip to Content
Merck
All Photos(1)

Key Documents

A7089

Sigma-Aldrich

Accumax solution

sterile-filtered, suitable for cell culture

Synonym(s):

Cell detachment solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

form

solution

concentration

1 ×

technique(s)

cell culture | mammalian: suitable
single cell analysis: suitable

pH

6.8-7.8

shipped in

dry ice

storage temp.

−20°C

Application

Accumax has proven effective in dissociating clumped cells in suspension cultures of hybridomas, CHO, BHK, 293, COS and Sf9 cells. The solution performed extremely well in dissociating cell counting, viral transfection assays, cell sorting, flow cytometry, and bioreactor scale-up. It has also been used to create single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue, and the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware.
TM=["Accumax"] solution has been used:
  • for detaching single walled carbon nanotubes from the extracellular matrix (ECM) in epithelial cells
  • for dissociation of retinal pigmented epithelial cells prior to passaging
  • for dissociation of islets from pancreas

Biochem/physiol Actions

Accumax is a cell detachment solution consisting of proteolytic and collagenolytic enzymes. Each bottle has 100 mL of 1x Accumax enzymes in Dulbecco′s PBS. There are no mammalian or bacterial-derived products found in Accumax.

Features and Benefits

Ready-to-use sterile liquid for in vitro cell applications

Caution

Accumax is stable when stored at -20°C. After thawing, the product can be stored for up to 2 months at 4°C.

Formula variant

Prepared in Dulbecco′s PBS (0.2 g/L KCl, 0.2 g/L KH2PO4, 8 g/L NaCl, and 1.15 g/L Na2HPO4). Refer to Product Data Sheet on the website for product usage information.

Preparation Note

This product is supplied as a sterile, ready-to-use frozen liquid. Accumax should be thawed at 37°C before use, and added at equal volume to a sample of clumped cells. Allow 5-10 minutes at 37°C for incubation and count cells as you would normally.

Legal Information

Accumax is a trademark of Innovative Cell Technologies, Inc.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of single walled carbon nanotubes at subtoxical concentrations on cell adhesion and other cell parameters of human epithelial cells
Kaiser JP, et al.
Journal of King Saud University - Science, 25(1), 15-27 (2013)
Natalja Funk et al.
Scientific reports, 9(1), 4515-4515 (2019-03-16)
Mutations within Leucine-rich repeat kinase 2 (LRRK2) are associated with late-onset Parkinson's disease. The physiological function of LRRK2 and molecular mechanism underlying the pathogenic role of LRRK2 mutations remain uncertain. Here, we investigated the role of LRRK2 in intracellular signal
Chronic beta-cell depolarization impairs beta-cell identity by disrupting a network of ca2+-regulated genes
Stancill JS, et al.
Diabetes, 66(8), 2175-2187 (2017)
Generation of retinal pigmented epithelium from iPSCs derived from the conjunctiva of donors with and without age related macular degeneration
Geng Z, et al.
PLoS ONE, 12(3), e0173575-e0173575 (2017)
Benjamin T Dodsworth et al.
Scientific reports, 10(1), 10542-10542 (2020-07-01)
Naïve human pluripotent stem cells (hPSC) resemble the embryonic epiblast at an earlier time-point in development than conventional, 'primed' hPSC. We present a comprehensive miRNA profiling of naïve-to-primed transition in hPSC, a process recapitulating aspects of early in vivo embryogenesis.

Articles

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

Lung organoids are valuable 3D models for human lung development and respiratory diseases. The 3dGRO™ differentiation protocol generates organoids from human iPSCs in 4 steps.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service