Skip to Content
Merck
All Photos(2)

Documents

A5879

Sigma-Aldrich

2-(2-Aminoethyl)isothiourea dihydrobromide

Synonym(s):

2-(2-Aminoethyl)-2-thiopseudourea dihydrobromide, AET, S-(2-Aminoethyl)isothiouronium bromide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SC(=NH)NH2 · 2HBr
CAS Number:
Molecular Weight:
281.01
Beilstein:
3911163
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (TLC)

form

powder

mp

190-196 °C (lit.)

solubility

water: 50 mg/mL, clear to very slightly hazy, colorless to yellow

SMILES string

Br[H].Br[H].NCCSC(N)=N

InChI

1S/C3H9N3S.2BrH/c4-1-2-7-3(5)6;;/h1-2,4H2,(H3,5,6);2*1H

InChI key

XDVMCVGTDUKDHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(2-Aminoethyl)isothiourea dihydrobromide inhibits nitric oxide synthase (NOS).

Biochem/physiol Actions

Inhibits constitutive and inducible NOS.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y Gundersen et al.
The European journal of surgery = Acta chirurgica, 165(12), 1167-1174 (2000-01-15)
To examine the effects of the inducible nitric oxide synthase inhibitor aminoethyl-isothiourea (AE-ITU) on haemodynamic measurements, and correlate these with hepatic morphology and function in a porcine model of endotoxaemia. Experimental study. 15 juvenile pigs. Flow probes were placed around
A Kis et al.
Journal of molecular and cellular cardiology, 31(6), 1229-1241 (1999-06-18)
Right ventricular pacing in lightly anaesthetized dogs (4x5 min periods at a pacing rate of 220 beats/min) protects against the consequences of coronary artery occlusion when this is initiated 24 h after the pacing stimulus. The main purpose of the
Meghan R Flanagan et al.
Annals of surgical oncology, 22(10), 3264-3272 (2015-07-24)
Adjuvant endocrine therapy (AET) has been shown to reduce the risk of second breast cancer events in women with ductal carcinoma in situ (DCIS). There is no population-level evaluation of AET use in DCIS patients after standardized reporting of estrogen
G J Southan et al.
British journal of pharmacology, 114(2), 510-516 (1995-01-01)
1. The induction of a calcium-independent isoform of nitric oxide (NO) synthase (iNOS) and a subsequent enhanced formation of NO has been implicated in the pathophysiology of a variety of diseases including inflammation and circulatory shock. Here we demonstrate that
Ni Ni Tun et al.
Plant & cell physiology, 47(3), 346-354 (2006-01-18)
In this study, we examined the regulation by putrescine, spermidine and spermine of nitric oxide (NO) biosynthesis in Arabidopsis thaliana seedlings. Using a fluorimetric method employing the cell-impermeable NO-binding dye diaminorhodamine-4M (DAR-4M), we observed that the polyamines (PAs) spermidine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service