Skip to Content
Merck
All Photos(3)

Key Documents

A5213

Sigma-Aldrich

Anti-β-Amyloid antibody, Mouse monoclonal

enhanced validation

clone BAM-10, ascites fluid

Synonym(s):

Anti-β-Amyloid antibody, Mouse monoclonal, Anti-A-BETA, Anti-Amyloid β Precursor Protein, Clone BAM91

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BAM-10, monoclonal

contains

15 mM sodium azide

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:2,000 using formic acid-treated, formalin-fixed, human Alzheimer′s disease (AD) brain sections.
indirect ELISA: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351) , APP(351)

Looking for similar products? Visit Product Comparison Guide

General description

β-amyloid protein or aβ4 is derived from larger protein that belongs to the family of 70kDa transmembrane glycoproteins (amyloid precursor proteins, APP). These are produced in various isoforms by alternative splicing. APPs are synthesized by many tissues including brain cells. Abnormal β-amyloid protein deposits have been associated with Alzheimer′s disease, Down′s syndrome, Dutch-type amyloidosis and Lewy body dementia.
The antibody reacts specifically with β-amyloid protein. The epitope recognized by the antibody resides within amino acids 1-12 of the β-amyloid protein. It specifically stains amyloid plaques within the cortex and amyloid deposits in blood vessels using formic acid-treated, formalin-fixed, paraffin-embedded, and Methacarn-fixed sections of human Alzheimer′s disease (AD) brain tissue.

Specificity

Monoclonal Anti-β-Amyloid Protein reacts specifically with β-amyloid protein. The epitope recognized by the antibody resides within amino acid residues 1-12 of the β-amyloid protein. The antibody specifically stains amyloid plaques within the cortex, and amyloid deposits in blood vessels, in formic acid-treated, formalin-fixed, paraffin-embedded and Methacarn-fixed sections of human Alzheimer′s disease (AD) brain tissue.

Immunogen

Synthetic β-amyloid peptide, conjugated to KLH.

Application

Monoclonal Anti- β Amyloid Protein may be used for the localization of β -amyloid protein using various immunochemical assays such as ELISA, competitive ELISA and immunohistochemistry.
Mouse monoclonal anti-ABETA was used to treat old WT PDAPP mice with amyloid accumulation and learning deficits in an attempt to improve learning and decrease accumulation, however no response was observed.
The antibody is useful in immunohistochemistry, immunoblotting, ELISA, and competitive ELISA. Also, this antibody has been used to neutralize Aβ assemblies in brains of transgenic mice expressing a mutant form of amyloid precursor protein, and for in vivo deep tissue imaging using near-IR optical spectrum.

Biochem/physiol Actions

β-amyloid fragments are amyloidogenic and neurotoxic both in vitro and in vivo. The presence of a large number of neuritic (senile) plaques and neurofibrillary tangles in the cerebral cortex is used as a pathological marker for a disease state and presents the major criterion for the diagnosis of Alzheimer′s disease at autopsy. A monoclonal antibody reacting specifically with β-amyloid protein is valuable for studying the nature of the β-amyloid protein by enabling detection and localization of β-amyloid protein and fragments.

Physical form

Monoclonal Anti-β-Amyloid Protein is provided as ascites fluid with 15mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for no more than one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tian Tian et al.
Nutrients, 8(9) (2016-09-13)
Alzheimer's disease (AD) is the most common type of dementia. Amyloid-β protein (Aβ) is identified as the core protein of neuritic plaques. Aβ is generated by the sequential cleavage of the amyloid precursor protein (APP) via the APP cleaving enzyme
Jared Ehrhart et al.
Journal of neuroinflammation, 2, 29-29 (2005-12-14)
Activated microglial cells have been implicated in a number of neurodegenerative disorders, including Alzheimer's disease (AD), multiple sclerosis (MS), and HIV dementia. It is well known that inflammatory mediators such as nitric oxide (NO), cytokines, and chemokines play an important
Jessica F Jordão et al.
PloS one, 5(5), e10549-e10549 (2010-05-21)
Immunotherapy for Alzheimer's disease (AD) relies on antibodies directed against toxic amyloid-beta peptide (Abeta), which circulate in the bloodstream and remove Abeta from the brain. In mouse models of AD, the administration of anti-Abeta antibodies directly into the brain, in
Folic acid deficiency enhances aβ accumulation in APP/PS1 mice brain and decreases amyloid-associated miRNAs expression
Liu, H, et al.
The Journal of Nutritional Biochemistry, 26(12), 1502-1508 (2015)
Kendra L Puig et al.
Neurobiology of aging, 40, 22-40 (2016-03-15)
APP/PS1 double transgenic mice expressing human mutant amyloid precursor protein (APP) and presenilin-1 (PS1) demonstrate robust brain amyloid beta (Aβ) peptide containing plaque deposition, increased markers of oxidative stress, behavioral dysfunction, and proinflammatory gliosis. On the other hand, lack of

Questions

  1. Please let us know the specificity of this antibody for Anti-beta Amyloid 1-40 or 1-41 or 1-42?

    1 answer
    1. The immunogen used to make this antibody corresponding to amino acids 1-40 of beta Amyloid. The antibody reacts specifically with β-amyloid protein. The epitope recognized by the antibody resides within amino acids 1-12 of the β-amyloid protein.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service