Skip to Content
Merck
All Photos(1)

Key Documents

A3734

Sigma-Aldrich

1-Azakenpaullone

≥97% (HPLC)

Synonym(s):

9-Bromo-7,12-dihydro-pyrido[3′,2′:2,3]azepino[4,5-b]indol-6(5H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrN3O
CAS Number:
Molecular Weight:
328.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

protect from light

color

gray-brown

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

O=C1CC2=C(NC3=C2C=C(Br)C=C3)C4=C(C=CC=N4)N1

InChI

1S/C15H10BrN3O/c16-8-3-4-11-9(6-8)10-7-13(20)18-12-2-1-5-17-15(12)14(10)19-11/h1-6,19H,7H2,(H,18,20)

InChI key

NTSBZVCEIVPKBJ-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

1-Azakenpaullone (AZP) can stimulate the transcription of β-catenin-related genes. This compound can induce the multiplication and replication of β cell.
1-Azakenpaullone is a cell permeable, potent, selective inhibitor of glycogen synthase kinase 3β (GSK3β) with 100-fold less cross-reactivity against CDKs.
1-Azakenpaullone is a cell permeable, potent, selective inhibitor of glycogen synthase kinase 3β (GSK3β) with 100-fold less cross-reactivity against CDKs. 1-Azakenpaullone can be used together with BIO and CHIR99021 as "gold standards" for GSK3 inhibition.

Caution

air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Hu et al.
Stem cells translational medicine, 6(1), 121-130 (2017-02-09)
Allogeneic hematopoietic stem cell transplantation (HSCT) is a potentially curative therapy for many malignant and nonmalignant diseases. However, chronic graft-versus-host disease (cGVHD) remains a significant cause of late morbidity and mortality after allogeneic HSCT. cGVHD often manifests as autoimmune syndrome.
Combination effects of AHR agonists and Wnt beta-catenin modulators in zebrafish embryos: implications for physiological and toxicological AHR functions
Wincent E, et al.
Toxicology and Applied Pharmacology, 284(2), 163-179 (2015)
9-cyano-1-azapaullone (cazpaullone), a glycogen synthase kinase-3 (GSK-3) inhibitor activating pancreatic beta cell protection and replication
Stukenbrock H, et al.
Journal of medicinal chemistry, 51(7), 2196-2207 (2008)
Thiranut Jaroonwitchawan et al.
Biochemical and biophysical research communications, 481(1-2), 176-181 (2016-11-07)
Neural crest (NC) is a transient population, arising during embryonic development and capable of differentiating into various somatic cells. The defects of neural crest development leads to neurocristopathy. Several signaling pathways were revealed their significance in NC cell specification. Fibroblast
Jens H Fritzenwanker et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(17), 8403-8408 (2019-04-11)
The trunk is a key feature of the bilaterian body plan. Despite spectacular morphological diversity in bilaterian trunk anatomies, most insights into trunk development are from segmented taxa, namely arthropods and chordates. Mechanisms of posterior axis elongation (PAE) and segmentation

Articles

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of serine/threonine kinases for over 100 proteins in many pathways. See a list of accepted GSK-3 modulators and related products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service