Skip to Content
Merck
All Photos(1)

Documents

A3275

Sigma-Aldrich

Anti-Human IgM (μ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Goat Anti-Human IgM (μ-chain specific)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

direct ELISA: 1:7,000-1:21,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgM antibody is produced by B cells and has pentamer structure which helps the antibody in polyreactivity and can also remove apoptotic cells. Anti-human IgM (μ-chain specific) -alkaline phosphatase antibody can be used in solid phase ELISA for determination of anti-sp75 antibodies. Goat anti human IgM-alkaline phosphatase antibody reacts specifically with human IgM.

Immunogen

Human IgM.

Application

IgM is a glycoprotein with 5 n-linked glycosylation sites on the heavy chain. An ELISA assay was performed to identify glycosylated forms of IgM that bind to lectin. Alkaline phosphatase conjugated goat anti-human IgM was used as the secondary at 1:2000 and developed using p-nitrophenyl substrate (Sigma).
It may be used for immunoblotting.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 15 mM sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tzer Chyn Lim et al.
Current biology : CB, 28(6), 955-962 (2018-03-06)
The position of the division site dictates the size and fate of daughter cells in many organisms. In animal cells, division-site placement involves overlapping mechanisms, including signaling from the central spindle microtubules, astral microtubules, and spindle poles and through polar
Immunoglobulin M Reactivity Towards the Immunologicdly Active Region sp75 of the Core Protein of Hepatitis C Virus (HCV) in Chronic HCV Infection.
U.B. Hellstrorn, S.P.E. Sylvan
Journal of Medical Virology, 39(4), 39325-39332 (1993)
Mandar Patgaonkar et al.
Parasite immunology, 40(10), e12580-e12580 (2018-08-14)
B cell-mediated humoral responses are essential for controlling malarial infection. Studies have addressed the effects of Plasmodium falciparum infection on peripheral B-cell subsets but not much is known for P. vivax infection. Furthermore, majority of the studies investigate changes during acute
Songwang Hou et al.
Journal of immunological methods, 440, 27-34 (2016-10-30)
The presence of circulating autoantibodies against phosphatidylethanolamine (PE) has been shown to be positively associated with symptoms of antiphospholipid syndromes (APS). However, the current ELISA-based tests for antiphosphatidylethanolamine (aPE) antibodies remain inconsistent and controversial. The term PE refers to a
Julia Whitehouse et al.
Vaccine, 34(15), 1792-1799 (2016-02-28)
Staphylococcus aureus is a pathogen which causes life-threatening infection, the incidence of which rises during adult life. This, together with the emergence of drug-resistant strains and the expansion of more susceptible elderly populations, represents the rationale for the ongoing development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service