Skip to Content
Merck
All Photos(3)

Key Documents

A3013

Sigma-Aldrich

Adenosine 2′(3′)-monophosphate mixed isomers

Synonym(s):

Yeast adenylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O7P
CAS Number:
Molecular Weight:
347.22
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Assay

≥45% (Adenosine-3′-monophosphate, HPLC)
≥97% (Total Adenosine-3′-monophosphate; Adenosine-2′-monophosphate, HPLC)
10-55% ( Adenosine-2′-monophosphate, HPLC)

form

powder

solubility

1 M NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](OP(O)(O)=O)[C@H]3O.Nc4ncnc5n(cnc45)[C@@H]6O[C@H](CO)[C@@H](O)[C@H]6OP(O)(O)=O

InChI

1S/2C10H14N5O7P/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(17)7(4(1-16)21-10)22-23(18,19)20;11-8-5-9(13-2-12-8)15(3-14-5)10-7(22-23(18,19)20)6(17)4(1-16)21-10/h2*2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20)/t2*4-,6-,7-,10-/m11/s1

InChI key

UQRXDDDXDPEXNS-VQFZJOCSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Adenosine 3′-monophosphate (3′-AMP) and Adenosine 2′-monophosphate (2′-AMP) are metabolites produced from hydrolysis of 2′,3′-cAMP by a family of metal-dependent phosphodiesterases. 2′-AMP and 3′-AMP inhibit proliferation of preglomerular vascular smooth muscle cells and glomerular mesangial cells via A2B receptors.

Application

Adenosine2′(3′)-monophosphate mixed isomers have been used as a standard to separate andquantify purines (adenine and guanine) and adenine ribonucleosidemonophosphates (2’-AMP and 3’-AMP) through isocratic high-performance liquidchromatography (HPLC). It has also been used to screen the multiplesugar-binding ATPase A (MsbA) transporter protein.

Linkage

Do not confuse with adenosine 2′:3′-cyclic monophosphate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Periprocedural type IVa myocardial infarction and the importance of platelet inhibition.
Anoop S V Shah et al.
Heart (British Cardiac Society), 99(17), 1225-1226 (2013-04-18)
Maria V Papadopoulou et al.
Oncology research, 14(1), 21-29 (2003-10-14)
The enzymatic cell-free metabolism of the novel hypoxia-selective cytotoxin 4-[3-(2-nitro-1-imidazolyl)-propylamino]-7-chloroquinoline hydrochloride (NLCQ-1) was investigated under hypoxic or aerobic conditions in the presence of purified reductive enzymes or isolated rat liver microsomes by monitoring the parent compound with HPLC-UV analysis. Enzymatic
G S Padiyar et al.
Journal of biomolecular structure & dynamics, 15(4), 803-821 (1998-03-26)
Crystal structures of lithium, sodium, potassium, calcium and magnesium salts of adenosine 2'-monophosphate (2'-AMP) have been obtained at atomic resolution by X-ray crystallographic methods. 2'-AMP.Li belongs to the monoclinic space group P2(1) with a = 7.472(3)A, b = 26.853(6) A
Alexander F Yakunin et al.
The Journal of biological chemistry, 279(35), 36819-36827 (2004-06-24)
In all mature tRNAs, the 3'-terminal CCA sequence is synthesized or repaired by a template-independent nucleotidyltransferase (ATP(CTP):tRNA nucleotidyltransferase; EC 2.7.7.25). The Escherichia coli enzyme comprises two domains: an N-terminal domain containing the nucleotidyltransferase activity and an uncharacterized C-terminal HD domain.
G S Padiyar et al.
Journal of biomolecular structure & dynamics, 15(4), 793-802 (1998-03-26)
This paper reports the crystal structures of free acid and ammonium salt of adenosine 2'-monophosphate (2'-AMP). 2'-AMP crystallizes in the hexagonal space group P6(5)22 with a = 9.530(3) A, c = 73.422(2) A, and Z= 12. 2'-AMP.NH4 crystallizes in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service