Skip to Content
Merck
All Photos(1)

Key Documents

A2835

Sigma-Aldrich

Amyloid Protein Non-Aβ Component

≥80% (HPLC)

Synonym(s):

NAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C141H235N39O49
CAS Number:
Molecular Weight:
3260.61
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥80% (HPLC)

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CO)NC(=O)CNC(=O)[C@H](C)NC(=O)CNC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)CNC(=O)CNC(=O)[C@@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](N)CCC(O)=O)C(C)C)[C@@H](C)O)C(C)C)C(C)C)C(C)C)[C@@H](C)O)C(C)C)[C@@H](C)O)C(C)C)[C@@H](C)O)C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H]([C@@H](C)O)C(=O)NCC(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](C(C)C)C(O)=O

InChI

1S/C141H235N39O49/c1-31-66(18)107(137(224)158-70(22)115(202)155-69(21)114(201)156-72(24)119(206)176-108(74(26)182)129(216)151-55-94(193)160-85(49-79-37-33-32-34-38-79)125(212)174-106(65(16)17)141(228)229)175-127(214)87(57-181)161-95(194)54-148-113(200)67(19)153-93(192)53-149-121(208)82(43-47-98(198)199)165-132(219)102(61(8)9)173-140(227)112(78(30)186)177-123(210)81(39-35-36-48-142)164-122(209)83(41-44-88(144)187)162-116(203)71(23)157-131(218)101(60(6)7)168-118(205)73(25)159-138(225)110(76(28)184)179-133(220)100(59(4)5)167-96(195)56-152-130(217)109(75(27)183)178-136(223)105(64(14)15)172-134(221)103(62(10)11)169-117(204)68(20)154-92(191)52-147-91(190)51-150-128(215)99(58(2)3)170-126(213)86(50-90(146)189)166-139(226)111(77(29)185)180-135(222)104(63(12)13)171-124(211)84(42-45-89(145)188)163-120(207)80(143)40-46-97(196)197/h32-34,37-38,58-78,80-87,99-112,181-186H,31,35-36,39-57,142-143H2,1-30H3,(H2,144,187)(H2,145,188)(H2,146,189)(H,147,190)(H,148,200)(H,149,208)(H,150,215)(H,151,216)(H,152,217)(H,153,192)(H,154,191)(H,155,202)(H,156,201)(H,157,218)(H,158,224)(H,159,225)(H,160,193)(H,161,194)(H,162,203)(H,163,207)(H,164,209)(H,165,219)(H,166,226)(H,167,195)(H,168,205)(H,169,204)(H,170,213)(H,171,211)(H,172,221)(H,173,227)(H,174,212)(H,175,214)(H,176,206)(H,177,210)(H,178,223)(H,179,220)(H,180,222)(H,196,197)(H,198,199)(H,228,229)/t66-,67-,68-,69-,70-,71-,72-,73-,74+,75+,76+,77+,78+,80-,81-,82-,83-,84-,85-,86-,87-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,111-,112-/m0/s1

InChI key

GXGKRPSFRUFLPR-PKXGUHEISA-N

Gene Information

human ... SNCA(6622)

Amino Acid Sequence

Glu-Gln-Val-Thr-Asn-Val-Gly-Gly-Ala-Val-Val-Thr-Gly-Val-Thr-Ala-Val-Ala-Gln-Lys-Thr-Val-Glu-Gly-Ala-Gly-Ser-Ile-Ala-Ala-Ala-Thr-Gly-Phe-Val

Biochem/physiol Actions

Found in the insoluble core of amyloid plaque; forms fibrils via a nucleation-dependent polymerization mechanism

Other Notes

Fragment 61-95 of a 140-amino acid amyloid precursor known as NACP, or α-synuclein

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congrong Wang et al.
Aging, 11(15), 5726-5743 (2019-08-14)
Carbohydrate overconsumption increases blood glucose levels, which contributes to the development of various diseases including obesity and diabetes. It is generally believed that high glucose metabolism increases cellular reactive oxygen species (ROS) levels, damages insulin-secreting cells and leads to age-associated
Haoli Yu et al.
Journal of biochemical and molecular toxicology, 33(8), e22352-e22352 (2019-06-04)
Cynaroside is a flavonoid compound proved to possess antioxidant activity, but its protective effect on age-related macular degeneration still remains unclear. In this study, the protective effects of cynaroside on oxidative stress and apoptosis in retinal pigment epithelial (RPE) cells
Jialun Li et al.
Theranostics, 9(20), 5839-5853 (2019-09-20)
Wound dressings composed of natural polymers, such as type I collagen, possess good biocompatibility, water holding capacity, air permeability, and degradability, and can be used in wound repair. However, due to the persistent oxidative stress in the wound area, the
Xiangdong Wang et al.
International journal of radiation biology, 95(8), 1051-1057 (2019-04-10)
Purpose: To investigate the relationship between autophagy and radiation damage of human hepatoma cells and to explore the role of reactive oxygen species (ROS). Materials and methods: HepG2 cells were exposed to X-rays, then the protein expressions of microtubule-associated protein
Hung-Chi Tu et al.
Ecotoxicology and environmental safety, 182, 109380-109380 (2019-07-07)
Ultraviolet (UV) is an omnipresent environmental carcinogen transmitted by sunlight. Excessive UV irradiation has been correlated to an increased risk of skin cancers. UVB, the most mutagenic component among the three UV constituents, causes damage mainly through inducing DNA damage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service