Skip to Content
Merck
All Photos(4)

Key Documents

A2179

Sigma-Aldrich

Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fab specific)–AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

should not react with

horse, human, bovine

technique(s)

direct ELISA: 1:40,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25
western blot (chemiluminescent): 1:80,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulins (Igs) belongs to the immunoglobulin super-family. Each immunoglobin have two heavy (H) and two light (L) chains held together by disulfide linkages.

Immunogen

Purified mouse IgG, Fab fragment

Application

Alkaline phosphatase-conjugated goat anti-mouse IgG (Fab specific) was used as the secondary antibody in ELISA assays to measure the reacitivity of mABs to thyrogobulin peptides. Absorbance was measured at 405 nm.
Anti-Mouse IgG (Fab specific)-Alkaline Phosphatase antibody produced in goat has been used in:
  • western blotting
  • immunoblotting
  • enzyme linked immunosorbent assay (ELISA)
  • immunohistochemistry

Biochem/physiol Actions

Digestion of IgG by papain results in generation of fragment antigen binding (Fab) comprising of one complete L chain and a variable and CH1 region of H chain. Pepsin digestion of IgG results in fragment crystallisable (fc), comprises of the H chain constant region. IgG antibody have enormous therapeutic potential and the Fc are is for the development of therapeutic antibody. Although the antibody site is located in the terminal end of the human IgG molecule (part of the Fab fragment), the Fc portion has various important functions such as complement fixation, site for rheumatoid factor (autoantibodies directed to Fc), passage through placental membrane and Staphylococcus protein A binding.
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice .

Other Notes

Antibody adsorbed with bovine, equine and human serum proteins.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with bovine, horse, or human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laszlo L P Hosszu et al.
Communications biology, 3(1), 402-402 (2020-07-31)
Prion diseases, a group of incurable, lethal neurodegenerative disorders of mammals including humans, are caused by prions, assemblies of misfolded host prion protein (PrP). A single point mutation (G127V) in human PrP prevents prion disease, however the structural basis for
Szymon W Manka et al.
Nature chemical biology (2023-01-17)
Recent cryogenic electron microscopy (cryo-EM) studies of infectious, ex vivo, prion fibrils from hamster 263K and mouse RML prion strains revealed a similar, parallel in-register intermolecular β-sheet (PIRIBS) amyloid architecture. Rungs of the fibrils are composed of individual prion protein
Szymon W Manka et al.
Nature communications, 13(1), 4004-4004 (2022-07-14)
Mammalian prions propagate as distinct strains and are composed of multichain assemblies of misfolded host-encoded prion protein (PrP). Here, we present a near-atomic resolution cryo-EM structure of PrP fibrils present in highly infectious prion rod preparations isolated from the brains
Increased expression of nuclear factor-kappaB in bronchial biopsies from smokers and patients with COPD
Di Stefano A, et al.
The European Respiratory Journal, 20(3), 556-563 (2002)
A Di Stefano et al.
The European respiratory journal, 20(3), 556-563 (2002-10-03)
The expression of nuclear factor (NF)-kappaB is an indicator of cellular activation and of inflammatory mediator production. The aim of the present study was to characterise the expression and localisation of p65, the major subunit of NF-kappaB, in the bronchial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service