Skip to Content
Merck
All Photos(2)

Key Documents

A2126

Sigma-Aldrich

Acetyl-β-methylcholine bromide

≥99%

Synonym(s):

Methacholine bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(CH3)CH2N(CH3)3Br
CAS Number:
Molecular Weight:
240.14
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99%

form

powder

mp

147-149 °C (lit.)

storage temp.

−20°C

SMILES string

[Br-].CC(C[N+](C)(C)C)OC(C)=O

InChI

1S/C8H18NO2.BrH/c1-7(11-8(2)10)6-9(3,4)5;/h7H,6H2,1-5H3;1H/q+1;/p-1

InChI key

MMVPLEUBMWUYIB-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

Application

Acetyl-β-methylcholine bromide (methylcholine bromide, methacholine bromide, Mch), a muscarinic acetylcholine receptor agonist, may be used in studies that involve the indentification, characterization and mechanism of action of muscarinic acetylcholine receptors. Mch may be used as a reference material in procedures used to detect or quantify its presence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David C Hornigold et al.
British journal of pharmacology, 138(7), 1340-1350 (2003-04-25)
1. We have examined possible mechanisms of cross-talk between the G(q/11)-linked M(3) muscarinic acetylcholine (mACh) receptor and the G(i/o)-linked M(2) mACh receptor by stable receptor coexpression in Chinese hamster ovary (CHO) cells. A number of second messenger (cyclic AMP, Ins(1,4,5)P(3))
John T Fisher et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 18(6), 711-713 (2004-02-24)
The presence of multiple muscarinic acetylcholine receptor (mAChR) subtypes in the heart and lung, combined with the lack of mAChR subtype-selective ligands, have complicated the task of identifying the mAChR subtypes mediating cardiac slowing (bradycardia) and airway narrowing (bronchoconstriction) due
Anyue Yin et al.
British journal of pharmacology, 175(16), 3394-3406 (2018-06-03)
Development of combination therapies has received significant interest in recent years. Previously, a two-receptor one-transducer (2R-1T) model was proposed to characterize drug interactions with two receptors that lead to the same phenotypic response through a common transducer pathway. We applied
M J van der Schans et al.
Journal of chromatography. A, 735(1-2), 387-393 (1996-05-31)
Solutions of histamine and methacholine bromide in different matrices for diagnostic purposes were analyzed for stability and quality control using capillary electrophoresis. Histamine (2,[4-imidazolyl]ethylamine) [CAS No. 51-45-6] was determined using a 0.1 M Tris-borate buffer of pH 8.3 with 5.10(-5)
M Yamada et al.
Nature, 410(6825), 207-212 (2001-03-10)
Members of the muscarinic acetylcholine receptor family (M1-M5) have central roles in the regulation of many fundamental physiological functions. Identifying the specific receptor subtype(s) that mediate the diverse muscarinic actions of acetylcholine is of considerable therapeutic interest, but has proved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service