Skip to Content
Merck
All Photos(1)

Key Documents

A211200

Sigma-Aldrich

DMT-2′Fluoro-dA(bz) Phosphoramidite

Synonym(s):

DMT-2′Fluoro-dA(bz) Amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H51FN7O7P
Molecular Weight:
875.92
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

nucleoside profile

base: deoxyadenosine
base protecting group: benzoyl
2' protecting group: fluoro
5' protecting group: DMT
deprotection: fast/standard

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]([C@H](O1)COC(C2=CC=C(OC)C=C2)(C3=CC=C(OC)C=C3)C4=CC=CC=C4)[C@@H](F)[C@@H]1N5C6=NC=NC(NC(C7=CC=CC=C7)=O)=C6N=C5

InChI

1S/C47H51FN7O7P/c1-31(2)55(32(3)4)63(60-27-13-26-49)62-42-39(61-46(40(42)48)54-30-52-41-43(50-29-51-44(41)54)53-45(56)33-14-9-7-10-15-33)28-59-47(34-16-11-8-12-17-34,35-18-22-37(57-5)23-19-35)36-20-24-38(58-6)25-21-36/h7-12,14-25,29-32,39-40,42,46H,13,27-28H2,1-6H3,(H,50,51,53,56)/t39-,40-,42-,46-,63?/m1/s1

InChI key

VCCMVPDSLHFCBB-MSIRFHFKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′Fluoro-dA(bz) Phosphoramidite is a 2′Fluoro phosphoramidite that is used to synthesize oligonucleotides that are more thermally stable and provide increased nuclease resistance.

Its key features include,

  • Can be employed together with DNA or RNA phosphoramidites
  • Recommended deprotection conditions are 8 hours at 55 °C using concentrated ammonia solution, or with AMA for 10 minutes at 65 °C
  • Synthesis of 2′Fluoro oligonucleotides is similar to standard DNA synthesis but requires an elongated coupling time (recommended is 3 minutes compared to 90 seconds for DNA monomers)
  • Consistent lot-to-lot high purity and performance
  • Manufactured under a certified ISO 9001 quality system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service