Skip to Content
Merck
All Photos(1)

Documents

A1765

Sigma-Aldrich

S-Acetyl-coenzyme A synthetase from baker′s yeast (S. cerevisiae)

lyophilized powder, ≥3 units/mg protein

Synonym(s):

Acetate CoA ligase (AMP forming), Acetate thiokinase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.26

form

lyophilized powder

Quality Level

specific activity

≥3 units/mg protein

composition

Protein, 10-30% biuret

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

S-Acetyl-coenzyme A synthetase from baker′s yeast (S. cerevisiae) has been used in the synthesis of adenosine 5′-tetraphosphate and adenosine 5′-pentaphosphate.
S-Acetyl-coenzyme A synthetase may be used to study various metabolic pathways, such as glycolysis, gluconeogenesis, pyruvate metabolism and CO2 fixation. It may also be used in gene expression studies.

Biochem/physiol Actions

Acetyl-coenzyme A synthetase catalyzes the production of acetyl-CoA. It is involved in histone acetylation in the nucleus. It may be involved in the growth of nonfermentable carbon sources such as glycerol. Acetyl-coenzyme A synthetase is induced by acetate, acetaldehyde and ethanol .

Packaging

Package size based on protein content.

Unit Definition

One unit will form 1.0 μmole of S-acetyl coenzyme A from acetate, ATP, and coenzyme A per min at pH 7.5 at 37 °C.

Physical form

Lyophilized powder containing stabilizers as potassium phosphate, sucrose, and reduced glutathione

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio Renilla et al.
Applied microbiology and biotechnology, 93(5), 2109-2124 (2011-09-02)
Impairment of acetate production in Escherichia coli is crucial for the performance of many biotechnological processes. Aerobic production of acetate (or acetate overflow) results from changes in the expression of central metabolism genes. Acetyl-CoA synthetase scavenges extracellular acetate in glucose-limited
Saurabh Sahar et al.
The Journal of biological chemistry, 289(9), 6091-6097 (2014-01-16)
The circadian clock regulates a wide range of physiological and metabolic processes, and its disruption leads to metabolic disorders such as diabetes and obesity. Accumulating evidence reveals that the circadian clock regulates levels of metabolites that, in turn, may regulate
Sara Castaño-Cerezo et al.
Molecular microbiology, 82(5), 1110-1128 (2011-11-09)
Lysine acetylation is a well-established post-translational modification widely conserved and distributed in bacteria. Although multiple regulatory roles have been proved, little is known about its regulation. Here, we present evidence that the transcription of the Gcn5-like acetyltransferase YfiQ of Escherichia
Seunghoon Lee et al.
Eukaryotic cell, 10(8), 1043-1052 (2011-06-15)
Acetyl coenzyme A (acetyl-CoA) is a crucial metabolite for energy metabolism and biosynthetic pathways and is produced in various cellular compartments with spatial and temporal precision. Our previous study on ATP citrate lyase (ACL) in Gibberella zeae revealed that ACL-dependent
Adenosine 5'-tetraphosphate and adenosine 5'-pentaphosphate are synthesized by yeast acetyl coenzyme A synthetase.
Guranowski A, et al.
Journal of Bacteriology, 176(10), 2986-2990 (1994)

Articles

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Enzyme Reagent Coenzyme A (CoA, CoASH or HSCoA) is the key cofactor in first step of the TCA cycle, responsible for transferring the acetyl group from pyruvate oxidation to oxaloacetate yielding citrate. Available through Sigma-Aldrich online.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service