Skip to Content
Merck
All Photos(1)

Key Documents

A1661

Sigma-Aldrich

N-(6-Aminohexyl)-N-ethylisoluminol

Synonym(s):

6-(N-[6-Aminohexyl]-N-ethyl)amino-2,3-dihydro-1,4-phthalazine-1,4-dione, AHEI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N4O2
CAS Number:
Molecular Weight:
304.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

form

powder

solubility

glacial acetic acid: 50 mg/mL

storage temp.

2-8°C

SMILES string

CCN(CCCCCCN)c1ccc2C(=O)NNC(=O)c2c1

InChI

1S/C16H24N4O2/c1-2-20(10-6-4-3-5-9-17)12-7-8-13-14(11-12)16(22)19-18-15(13)21/h7-8,11H,2-6,9-10,17H2,1H3,(H,18,21)(H,19,22)

InChI key

BEADQYOTAORBKI-UHFFFAOYSA-N

Application

Chemiluminescence is dramatically increased (up to 15×) in the presence of γ-cyclodextrin, with implications for improved sensitivity of biochemical assays.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijuan Zhang et al.
Nutrition (Burbank, Los Angeles County, Calif.), 54, 12-18 (2018-04-29)
We examined the feasibility and acceptability of a non-restrictive diet that was focused on increasing dietary fiber and lean protein intake for weight loss. Dietary intake was assessed using three randomly selected 24-h dietary recalls. Fifteen obese adults enrolled in
Giuseppe Grosso et al.
International journal of environmental research and public health, 17(5) (2020-02-29)
Background: Current scientific literature suggests healthy dietary patterns may have less environmental impact than current consumption patterns, but most of the studies rely on theoretical modeling. The aim of this study was to assess the impact on resources (land, water
Andrea K Chomistek et al.
Journal of the American College of Cardiology, 65(1), 43-51 (2015-01-13)
Overall mortality rates from coronary heart disease (CHD) in the United States have declined in recent decades, but the rate has plateaued among younger women. The potential for further reductions in mortality rates among young women through changes in lifestyle
Nancy López-Olmedo et al.
Public health nutrition, 22(15), 2777-2792 (2019-06-14)
The present study evaluated the association of two measures of diet quality with BMI and waist circumference (WC), overall and by education level, among Mexican men and women. We constructed two a priori indices of diet quality, the Mexican Diet
Jiyoung Kim et al.
Clinical nutrition research, 2(2), 100-106 (2013-08-03)
The present study was performed to evaluate the relationship between dietary quality indices including the Diet Quality Index-International (DQI-I), Alternate Healthy Eating Index (AHEI), and Healthy Diet Indicator (HDI) and glycemic status in Korean patients with type 2 diabetes. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service