Skip to Content
Merck
All Photos(1)

Documents

A1332

Sigma-Aldrich

Ammonium citrate tribasic

≥97% (titration)

Synonym(s):

Citric acid triammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CO2NH4)(CH2CO2NH4)2
CAS Number:
Molecular Weight:
243.22
Beilstein:
3779194
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97% (titration)

form

crystals

bp

100 °C (lit.)

mp

185 °C (dec.) (lit.)

density

1 g/mL at 25 °C (lit.)

application(s)

diagnostic assay manufacturing

SMILES string

N.N.N.OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7.3H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*1H3

InChI key

YWYZEGXAUVWDED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Preparation of One-Emission Nitrogen-Fluorine-Doped Carbon Quantum Dots and Their Applications in Environmental Water Samples and Living Cells for ClO(-) Detection and Imaging.: This study illustrates the role of ammonium citrate tribasic in synthesizing nitrogen-fluorine-doped carbon quantum dots. These quantum dots are used for sensitive detection and imaging of chlorite ions in environmental water samples and living cells, demonstrating a novel application in environmental monitoring and biomedical imaging (Zhang et al., 2023).

  • High quantum yield nitrogen and boron co-doped carbon dots for sensing Ag(+), biological imaging and fluorescent inks.: Ammonium citrate tribasic is used here to facilitate the doping process in the synthesis of high quantum yield carbon dots. These dots have applications ranging from heavy metal ion sensing in environmental samples to advanced imaging techniques in biological systems and the creation of fluorescent inks (Cheng et al., 2021).

  • Optimization of Copper Stain Removal from Marble through the Formation of Cu(II) Complexes in Agar Gels.: This research employs ammonium citrate tribasic in a novel method for removing copper stains from marble. The study explores the chelating properties of ammonium citrate tribasic to form stable Cu(II) complexes, offering a conservation technique for historical artifacts and buildings (Sansonetti et al., 2021).

  • A facile microwave-assisted synthesis of highly crystalline red carbon dots by adjusting the reaction solvent for white light-emitting diodes.: Ammonium citrate tribasic is instrumental in this study for synthesizing red carbon dots under microwave conditions. The dots are used in the development of white light-emitting diodes, showing potential for applications in display technologies and lighting (Lan et al., 2020).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David Lubkowicz et al.
ACS synthetic biology, 7(5), 1229-1237 (2018-04-14)
Gram-positive Staphylococcus aureus infection that results in pneumonia, urinary tract infection, and in severe cases, sepsis, has recently been classified as a serious threat to public health. Rapid and cost-effective detection of these infections are costly and time-consuming. Here, we
Kiyohito Totsuka et al.
Experimental eye research, 181, 316-324 (2018-09-02)
The dysfunction and cell death of retinal pigment epithelial (RPE) cells are hallmarks of late-stage dry (atrophic) age-related macular degeneration (AMD), for which no effective therapy has yet been developed. Previous studies have indicated that iron accumulation is a source
Kouhei Mizuno et al.
Scientific reports, 7(1), 12880-12880 (2017-10-19)
In certain conditions, members of the Lactobacillus genus are auxotrophs that have fastidious requirements for growth. Notably, Lactobacillus cannot grow in M9 medium, a minimal synthetic medium used for Escherichia coli. However, we found that some Lactobacillus strains can be
Carolyn R O'Brien et al.
Journal of feline medicine and surgery, 19(5), 498-512 (2017-04-26)
This paper, the first in a series of three on 'feline leprosy', provides a detailed description of disease referable to Candidatus 'Mycobacterium tarwinense', the most common cause of feline leprosy in Victoria, Australia. Cases were sourced retrospectively and prospectively for
Jungnam Lee et al.
PloS one, 13(4), e0194803-e0194803 (2018-04-13)
Idiopathic pulmonary fibrosis (IPF) is a chronic lung disease of unknown cause characterized by alveolar epithelial damage, patchy interstitial fibrosis and diffuse microvascular abnormalities. In IPF, alveolar clustering of iron-laden alveolar macrophages-a common sign of microhemorrhage, has been associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service