Skip to Content
Merck

A1237

Pramipexole dihydrochloride

>98% (HPLC), Dopamine D2 and D3 receptor agonist, powder

Synonym(s):

(S)-2-amino-4,5,6,7-tetrahydro-6-(propylamino)benzothiazole dihydrochloride, PPX dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H17N3S · 2HCl
CAS Number:
Molecular Weight:
284.25
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
>98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pramipexole dihydrochloride, >98% (HPLC), powder

assay

>98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

H2O: >20 mg/mL

originator

Boehringer Ingelheim

storage temp.

2-8°C

SMILES string

Cl.Cl.CCCN[C@H]1CCc2nc(N)sc2C1

InChI

1S/C10H17N3S.2ClH/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;/h7,12H,2-6H2,1H3,(H2,11,13);2*1H/t7-;;/m0../s1

InChI key

QMNWXHSYPXQFSK-KLXURFKVSA-N

Gene Information

General description

Pramipexole belongs to non-ergoline class and is derived from benzothiazole. It has strong affinity for D2 and D3 receptors (dopamine receptors) compared to other ergoline-derived drugs. However, it does not associate with non-dopamine receptors, for instance adrenergic and serotonin receptors.

Application

Pramipexole dihydrochloride has been used as D2 receptor agonist in dopamine (DA)-depleted striatum slices.
Pramipexole dihydrochloride has been used as a D2-like DR (dopaminergic receptor) agonist in study associated with Parkinson′s disease. It has also been used as a D2/D3 receptor agonist to study risky decision-making in rats.

Biochem/physiol Actions

Pramipexole dihydrochloride monohydrate is a dopamine agonist active at D3 and D2 receptor subtypes.
Pramipexole elicits neuroprotective role and may help in treating depression. It also has a potential to modulate limb movement and may be effective in treating restless legs syndrome.
Pramipexole is a dopamine agonist active at D3 and D2 receptor subtypes. Pramipexole has been found to have neuroprotective effects independent of its dopamine receptor agonism. It reduces mitochondrial reactive oxygen species (ROS) production and inhibits the activation of apoptotic pathways.
Pramipexole is a therapeutic agent for Parkinson′s disease and restless leg syndrome. It also exhibits antidepressant responses in patients suffering from MDD (major depressive disorder) and bipolar depression.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Dopamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library