Skip to Content
Merck
All Photos(4)

Key Documents

A1009

Sigma-Aldrich

Albumin methylated from bovine serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201
NACRES:
NA.26

biological source

bovine

Quality Level

form

powder

origin

USA origin

technique(s)

affinity chromatography: suitable
fractionation: suitable

impurities

≤10% water (Karl Fischer)

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Looking for similar products? Visit Product Comparison Guide

Application

Albumin methylated from bovine serum (mBSA) has been used as a coating for immunoplates in enzyme-linked immunosorbent assay (ELISA) of anti-dsDNA IgG antibodies. It has also been used to induce arthritis in mice models.
As an affinity ligand on diatomaceous earth support (MAK chromatography), a medium for fractionation of different forms of DNA and for separating aminoacyl-tRNAs. Different modes of binding of double-stranded, denatured, and replicating forms of DNA account for the efficacy of the method.

Biochem/physiol Actions

Albumin is a major circulating component of the plasma and is a highly soluble multifunctional protein. It plays a role in several physiological and pharmacological processes. Albumin binds to metals, cholesterol, hormones, bile pigments, drugs, and fatty acids to facilitate their transport. It regulates osmotic pressure and distributes fluids between various compartments.

Preparation Note

Methyl ester of albumin prepared per Mandell, J.D. and Hershey, A.D., Anal. Biochem., 1, 66 (1960).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kasper Almholt et al.
Journal of immunology (Baltimore, Md. : 1950), 200(3), 957-965 (2017-12-29)
Genetic absence of the urokinase-type plasminogen activator (uPA) reduces arthritis progression in the collagen-induced arthritis (CIA) mouse model to an extent just shy of disease abrogation, but this remarkable observation has not been translated into therapeutic intervention. Our aim was
Wim B van den Berg et al.
Methods in molecular medicine, 136, 243-253 (2007-11-07)
Antigen induced arthritis is a unilateral T-cell driven model caused by direct injection of an antigen into the knee joint of a FCA preimmunized animal. The chronicity is determined by antigen retention in avascular structures of the joint through charge
Fractionation of nucleic acids with the methylated albumin column.
N SUEOKA et al.
Journal of molecular biology, 4, 161-172 (1962-03-01)
David L Mitchell
Methods in molecular biology (Clifton, N.J.), 314, 239-249 (2006-05-06)
Over the past 20 yr, the use of polyclonal and monoclonal antibodies to quantify damage in DNA has burgeoned. Immunoassays offer distinct advantages over other anaytical procedures currently used to measure DNA damage including adaptability, sensitivity and selectivity. This combination
T Hofstaetter et al.
Biochimica et biophysica acta, 518(1), 53-60 (1978-03-29)
The binding of methylated albumin to DNA, the basis of the chromatography on columns of kieselguhr coated with methylated albumin (MAK chromatography), was investigated. Scatchard plots revealed only one mode of interaction with fully double-stranded DNA. The complexes should be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service