Skip to Content
Merck
All Photos(1)

Key Documents

A0156

Sigma-Aldrich

N-(4-Aminobutyl)-N-ethylisoluminol

≥90%, powder

Synonym(s):

4-(N1-Ethyl-4-aminobutylamino)phthalic hydrazide, 6-[N-(4-Aminobutyl)-N-ethylamino]-2,3-dihydro-1,4-phthalazinedione, ABEI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N4O2
CAS Number:
Molecular Weight:
276.33
Beilstein:
920895
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%

Assay

≥90%

form

powder

mp

259-260 °C (lit.)

solubility

glacial acetic acid: 50 mg/mL, clear, colorless to yellow

storage temp.

2-8°C

SMILES string

CCN(CCCCN)c1ccc2C(=O)NNC(=O)c2c1

InChI

1S/C14H20N4O2/c1-2-18(8-4-3-7-15)10-5-6-11-12(9-10)14(20)17-16-13(11)19/h5-6,9H,2-4,7-8,15H2,1H3,(H,16,19)(H,17,20)

InChI key

LEOJISUPFSWNMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Efficient chemiluminescent NH2-coupling reagent for detection of a wide variety of proteins down to the picomole range.

Other Notes

Reportedly, the use of this material in chemiluminescent assays has distinct advantages over conventional radioimmunoassay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shulin Zhao et al.
Chemical communications (Cambridge, England), 48(5), 699-701 (2011-12-03)
We report on a novel strategy to improve microfluidic immunoassay sensitivity by introducing chemiluminescence resonance energy transfer (CRET) into the immunoreactions. The proposed CRET-based immunoassay for estradiol (E2, as a model analyte) is one of the most sensitive immunoassay with
Yating Liu et al.
Nature communications, 8(1), 1003-1003 (2017-10-19)
Most known chemiluminescence (CL) reactions exhibit flash-type light emission. Great efforts have been devoted to the development of CL systems that emit light with high intensity and long-lasting time. However, a long-lasting CL system that can last for hundreds of hours
Shulin Zhao et al.
Analytical biochemistry, 393(1), 105-110 (2009-06-23)
A microchip electrophoresis (MCE) method with chemiluminescence (CL) detection was developed for the determination of carnosine-related peptides, including carnosine, homocarnosine, and anserine, in biological samples. A simple integrated MCE-CL system was built to perform the assays. The highly sensitive CL
O M Steijger et al.
Journal of chromatography, 615(1), 97-110 (1993-05-19)
N-(4-Aminobutyl)-N-ethylisoluminol was used for labelling of carboxylic acids. The derivatization reaction was carried out with 1-hydroxybenzotriazole as pre-activator of the carboxylic acid function and N-ethyl-N'-(3-dimethylaminopropyl)carbodiimide as the coupling reagent. Optimum conditions for the derivatization were determined by using factorial design
Zhouping Wang et al.
Analytical and bioanalytical chemistry, 398(5), 2125-2132 (2010-09-14)
A highly selective electrochemiluminescent biosensor for the detection of target nephrotoxic toxin, ochratoxin A (OTA), was developed using a DNA aptamer as the recognition element and N-(4-aminobutyl)-N-ethylisoluminol (ABEI) as the signal-producing compound. The electrochemiluminescent aptamer biosensor was fabricated by immobilizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service