Recommended Products
form
oil
refractive index
n20/D 1.493-1.513
density
0.940-1.000 g/mL at 20 °C
0.952 g/mL at 25 °C (lit.)
lipid type
oils
Looking for similar products? Visit Product Comparison Guide
Application
- Cedar Oil in Hydrogel Composite Development: Cedar essential oil was integrated into polyvinyl alcohol/cedar oil/kaolin composite hydrogels to enhance hemostatic, antibacterial, and antioxidant properties. This application underscores its potential in biomedical material engineering, particularly in wound management products (Tamer et al., 2022).
- Food Packaging Applications: Cedar oil was utilized in the development of bioactive polypropylene films, demonstrating its efficacy as a natural preservative in food packaging. This aligns with the needs of the food industry to extend product shelf life while using sustainable, non-toxic materials (Antosik et al., 2021).
- Antioxidant and Antibacterial Activities: The characterization of cedarwood essential oil emulsions revealed significant antioxidant and antibacterial activities, suggesting its applicability in food safety and preservation sectors. This application highlights the oil′s dual functionality, which can be leveraged in various industrial manufacturing processes (Huang et al., 2021).
- Larvicidal Performance Enhancements: Cedarwood essential oil was effectively used in pectin-cedarwood essential oil nanocapsules, which were impregnated onto cotton bags to improve larvicidal performances against mosquito larvae. This indicates its potential use in public health applications for vector control (Kala et al., 2020).
Biochem/physiol Actions
Cedar oil demonstrates antimicrobial activity against anaerobic bacteria and yeast, as assessed by a standardized broth assay .
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - Skin Sens. 1B
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sibin Mathew Nesin et al.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association, 28(6), 1640-1653 (2019-03-25)
Stroke leads to devastating impact on health as well as quality of life making it one of the leading causes of disability. Restoring the functions of upper extremities after ischemic (ISC) stroke is one of the challenges for rehabilitation. Lack
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service