Skip to Content
Merck
All Photos(1)

Key Documents

94330

Sigma-Aldrich

Uridine 5′-diphosphate disodium salt hydrate

≥96.0% (HPLC)

Synonym(s):

UDP, 5′-UDP-Na2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2Na2O12P2 · xH2O
CAS Number:
Molecular Weight:
448.12 (anhydrous basis)
Beilstein:
8817400
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic

Assay

≥96.0% (HPLC)

form

powder

impurities

≤15% water

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP(O)([O-])=O)N2C=CC(=O)NC2=O

InChI

1S/C9H14N2O12P2.2Na.H2O/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,10,12,15)(H2,16,17,18);;;1H2/q;2*+1;/p-2/t4-,6-,7-,8-;;;/m1.../s1

InChI key

RHBXRWOXLRCWIZ-LLWADOMFSA-L

Application

Uridine 5′-diphosphate disodium salt hydrate has been used:
  • as a standard for quantification of metabolite levels in murine tumor interstitial fluid by liquid chromatography-mass spectrometry (LC/MS)
  • as a UDP standard for nucleotide analysis by liquid chromatography-high resolution mass spectrometry (LC-HRMS)
  • to treat E6.5 retina explants in vitro, to study its effect on the entry and elongation of microglial cells into the embryonic quail retina

Biochem/physiol Actions

Uridine 5′-diphosphate (UDP) is an endogenous signaling molecule produced by damaged cells to attract macrophages. In response to neuronal damage, UDP promotes chemotaxis and chemokinesis in microglial cells. UDP serves as a ligand for P2Y receptors. UDP and uridine 5′-triphosphate (UTP) may be used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled protein receptors (GPCR) and cell signaling molecules such as Rho-signaling via guanine nucleotide exchange factors (GEF).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenneth A Jacobson et al.
Purinergic signalling, 5(1), 75-89 (2008-07-05)
Although elucidation of the medicinal chemistry of agonists and antagonists of the P2Y receptors has lagged behind that of many other members of group A G protein-coupled receptors, detailed qualitative and quantitative structure-activity relationships (SARs) were recently constructed for several
María Martín-Estebané et al.
PloS one, 12(8), e0182450-e0182450 (2017-08-02)
Microglial cell precursors located in the area of the base of the pecten and the optic nerve head (BP/ONH) start to enter the retina of quail embryos at the 7th day of incubation (E7), subsequently colonizing the entire retina by
Caroline Liot et al.
PloS one, 6(8), e23676-e23676 (2011-09-03)
Besides regulation of actin cytoskeleton-dependent functions, Rho GTPase pathways are essential to cell cycle progression and cell division. Rho, Rac and Cdc42 regulate G1 to S phase progression and are involved in cytokinesis. RhoA GDP/GTP cycling is required for normal
Feng Ding et al.
Current topics in medicinal chemistry, 11(23), 2879-2887 (2011-08-10)
The Rho family small GTPases of the Ras superfamily play key roles in regulating diverse signaling pathways that control a myriad of fundamental cellular processes such as cytoskeletal dynamics, cell cycle progression, gene expression, cell polarity, migration and cell transformation.
Raquel Buj et al.
Cell reports, 28(8), 1971-1980 (2019-08-23)
Reprogrammed metabolism and cell cycle dysregulation are two cancer hallmarks. p16 is a cell cycle inhibitor and tumor suppressor that is upregulated during oncogene-induced senescence (OIS). Loss of p16 allows for uninhibited cell cycle progression, bypass of OIS, and tumorigenesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service