Skip to Content
Merck
All Photos(1)

Key Documents

93017

Sigma-Aldrich

SSC Buffer 20x

for Northern and Southern blotting, solution

Synonym(s):

20x SSC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

solution

technique(s)

Southern blotting: suitable

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

pH

7.0±0.2

density

1.16 g/mL at 20 °C

storage temp.

room temp

General description

SSC Buffer 20x Concentrate is a standard reagent in Southern and Northern blotting procedures. This concentrated formula can be used straight or diluted to make SSC working solutions with concentrations as low as 0.5x.

Application

Suitable for:
  • Preparing nucleic acids for hybridization
  • Hybridization buffer for Northern and Southern blots
  • Washing buffer for Northern and Southern blots

Features and Benefits

  • Convenient ready-to-use liquid
  • Multi-purpose use in hybridizations
  • Easily diluted to any concentration needed

Components

SSC Buffer 20x Concentrate contains 0.3 M sodium citrate in 3M NaCl.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanghua Yang et al.
BMC biology, 11, 86-86 (2013-07-23)
Human pluripotent stem cells (hPSCs) hold great promise for applications in regenerative medicine. However, the safety of cell therapy using differentiated hPSC derivatives must be improved through methods that will permit the transplantation of homogenous populations of a specific cell
Sterghios A Moschos et al.
BMC genomics, 8, 240-240 (2007-07-21)
At present, nothing is known of the role of miRNAs in the immune response in vivo despite the fact that inflammation is thought to underlie multiple acute and chronic diseases. In these circumstances, patients are commonly treated with corticosteroids such

Articles

Learn how water impurities can affect DNA and RNA electrophoresis and blotting. High purity water purified using ultrafiltration is suitable for nucleic acid electrophoresis, Northern and Southern blotting.

Learn how water impurities can affect DNA and RNA electrophoresis and blotting. High purity water purified using ultrafiltration is suitable for nucleic acid electrophoresis, Northern and Southern blotting.

Learn how water impurities can affect DNA and RNA electrophoresis and blotting. High purity water purified using ultrafiltration is suitable for nucleic acid electrophoresis, Northern and Southern blotting.

Learn how water impurities can affect DNA and RNA electrophoresis and blotting. High purity water purified using ultrafiltration is suitable for nucleic acid electrophoresis, Northern and Southern blotting.

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service