Skip to Content
Merck
All Photos(1)

Key Documents

86186

Sigma-Aldrich

Sulforhodamine B acid chloride

suitable for fluorescence, technical

Synonym(s):

Lissamine rhodamine B sulfonylchloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H29ClN2O6S2
CAS Number:
Molecular Weight:
577.11
Beilstein:
7734927
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

grade

technical

form

powder

fluorescence

λex 560 nm; λem 580 nm in chloroform
λex 560 nm; λem ~580 nm in chloroform

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCN(CC)c1ccc2c(OC3=C\C(C=CC3=C2c4ccc(cc4S([O-])(=O)=O)S(Cl)(=O)=O)=[N+](/CC)CC)c1

InChI

1S/C27H29ClN2O6S2/c1-5-29(6-2)18-9-12-21-24(15-18)36-25-16-19(30(7-3)8-4)10-13-22(25)27(21)23-14-11-20(37(28,31)32)17-26(23)38(33,34)35/h9-17H,5-8H2,1-4H3

InChI key

YERWMQJEYUIJBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulforhodamine B is a fluorescent dye.

Application

Fluorescent protein label forming stable conjugates
Sulforhodamine B acid chloride has been used to synthesize rhodamine-labeled polycation (PLL-ROD). It has also been used in fluorescent protein tracer preparation.

Biochem/physiol Actions

The sulforhodamine B (SRB) assay is useful in determining the density of cells.

Legal Information

Lissamine is a trademark of PerkinElmer Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A combined numerical and experimental framework for determining permeability properties of the arterial media
Comerford A, et al.
Biomechanics and Modeling in Mechanobiology, 14(2), 297-313 (2015)
Anticancer Activities of African Medicinal Spices and Vegetables
Medicinal Spices and Vegetables from Africa, 271-297 (2017)
J Rosenecker et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(14), 7236-7241 (1996-07-09)
We have used a pharmacologic mediator to open intercellular connections in selected vessels to allow liposomes to escape from the blood stream and to extravasate into tissues that have appropriate receptors. We have examined the effects of substance P (SP)
J W Sanger et al.
The Journal of cell biology, 99(3), 918-928 (1984-09-01)
To determine if a living cell is necessary for the incorporation of actin, alpha-actinin, and tropomyosin into the cytoskeleton, we have exposed cell models to fluorescently labeled contractile proteins. In this in vitro system, lissamine rhodamine-labeled actin bound to attachment
J M Kagan
Journal of dental research, 59, 1750-1756 (1980-11-01)
The histopathologic status of gingival tissue was assessed according to the criteria of Page and Schroeder.(5) Rhodamine-labeled bacteria were used to assess the antigenic specificity of plasma cells in the tissue. Examination of tissue sections revealed that 31.6% of plasma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service