Skip to Content
Merck
All Photos(1)

Key Documents

84120

Sigma-Aldrich

Safranin O

for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0)

Synonym(s):

Basic Red 2, Cotton Red, Gossypimine, Safranin T, Safranin Y or A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Colour Index Number:
50240
Beilstein:
3924099
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

for microscopy (Bact., Bot., Hist.)
indicator (pH 0.3-1.0)

Quality Level

form

powder or crystals

technique(s)

titration: suitable

pH

10 (20 °C, 10 g/L)

solubility

methanol: 0.01 g/10 mL, red

εmax

≥1000 at 525-535 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology
microbiology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

InChI

1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H

InChI key

OARRHUQTFTUEOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Safranin O, also known as Basic Red 2, is a basic, cationic, and metachromatic azine dye used as a biological stain. The characteristic azine chromophore contains the phenazine, a planar, tricyclic dibenzopyrazine, which consists of two non-planar benzene rings linked by two nitrogen atoms. A delocalized positive charge, an alternation of aromatic and quinonoid structures, and diazotizable amine groups constitute the chromophoric system. c-Methylation makes this dye of moderate overall size. The trimethyl cationic form of safranine is lipophilic, whereas the dimethyl forms are weakly hydrophilic. The dye is commercially available as a chloride.

Application

  • Safranin O is widely employed as a microbiological stain, especially as a counterstain in Gram staining to distinguish Gram-positive from Gram-negative micro-organisms, and for staining bacterial spores.
  • It is used to demonstrate glycosaminoglycans in human and animal histological specimens and nuclei.
  • It is a component of Benda′s polychrome stain.
  • It is a component of various polychromes and lignin stains for botanical staining applications.
  • It has also been applied for the demonstration of mast cells in cytospin preparations and staining surgical frozen sections.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Conn's Biological Stains
Kiernan & Horobin
Conn?s Biological Stains (2002)
José G Raya et al.
Investigative radiology, 46(6), 401-409 (2011-03-24)
To investigate changes of diffusion tensor imaging (DTI) parameters (mean apparent diffusion coefficient [ADC], fractional anisotropy [FA], and first eigenvector) with increasing proteoglycan (PG) extraction of articular cartilage. Twelve cylindrical cartilage-on-bone samples were drilled from 4 human patellae (3 per
Tian Gan et al.
Talanta, 85(1), 310-316 (2011-06-08)
A disposable electrochemical sensor for the determination of indole-3-acetic acid (IAA) based on nanocomposites of reduced graphene oxide (rGO) and poly(safranine T) (PST) was reported. The sensor was prepared by coating a rGO film on a pre-anodized graphite electrode (AGE)
Nina Orfali et al.
Molecular oncology, 14(6), 1297-1309 (2019-12-11)
Ubiquitin/ISG15-conjugating enzyme E2L6 (UBE2L6) is a critical enzyme in ISGylation, a post-translational protein modification that conjugates the ubiquitin-like modifier, interferon-stimulated gene 15 (ISG15), to target substrates. Previous gene expression studies in acute promyelocytic leukemia (APL) cells showed that all-trans-retinoic acid
Finely tuned temperature-controlled cargo release using paraffin-capped mesoporous silica nanoparticles.
Elena Aznar et al.
Angewandte Chemie (International ed. in English), 50(47), 11172-11175 (2011-09-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service