Skip to Content
Merck
All Photos(1)

Documents

76258

Sigma-Aldrich

Paraplast Plus®

suitable for electron microscopy, pellets (fast-melting)

Synonym(s):

Paraffin – polyisobutylene mixture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

pellets (fast-melting)

technique(s)

electron microscopy: suitable

thickness

≤1 μm , short ribbons
≤2 μm , sectioning: continuous ribbons

mp

56-57 °C

application(s)

hematology
histology

storage temp.

2-8°C

General description

regular Paraplast with approx. 0.8% DMSO added for enhanced ease of infiltration and sectioning

Application

Paraffin polyisobutylene mixture is a tissue embedding medium composed of highly purified paraffin and polyisobutylene and a low level of dimethyl sulfoxide (DMSO) to enhance the rate of tissue penetration. Paraffin-polyisobutylene (76258) supports thin wrinkle-free sectioning down to 4 microM with more flexible and continuous ribbons.

Legal Information

Paraplast Plus is a registered trademark of Leica Biosystems

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H O Goyal et al.
The Anatomical record, 250(2), 164-171 (1998-03-07)
Since androgens and/or estrogens must bind with specific receptors in order to elicit a response at the target organ(s), it is important to understand factors that regulate expression of androgen receptors (AR) and estrogen receptors (ER). Hence, the objective of
Ewa Urbańczyk-Wochniak et al.
Cellular & molecular biology letters, 7(1), 7-18 (2002-04-11)
This study describes an effective method of in situ RT-PCR (RT-ISPCR) that was developed to localize gene expression in plant tissues. This RT-PCR technique was performed on sectioned tissues of female buds of the cucumber GY3 inbred line. The CUS1
Ye Jin et al.
The Plant cell, 21(7), 2072-2089 (2009-07-04)
Invertase plays multiple pivotal roles in plant development. Thus, its activity must be tightly regulated in vivo. Emerging evidence suggests that a group of small proteins that inhibit invertase activity in vitro appears to exist in a wide variety of
R S Liem et al.
Acta anatomica, 132(2), 164-168 (1988-01-01)
A method is described for the histochemical detection of horseradish peroxidase in Paraplast Plus embedded brain sections. The procedure uses 150-micron-thick Vibratome-cut slices of glutaraldehyde-paraformaldehyde-fixed brain tissue. Tetramethylbenzidine stabilized by diaminobenzidine/cobalt/H2O2 is used as chromogen. The Vibratome-cut slices are dehydrated
[Paraplast-plus, a tissue embedding medium that enables sections to stick to the slides without adhesives (author's transl)].
D Arn et al.
Mikroskopie, 31(3-4), 99-106 (1975-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service