Skip to Content
Merck
All Photos(1)

Key Documents

69890

Sigma-Aldrich

MES hydrate

BioUltra, ≥99.5% (T)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid hydrate, 4-Morpholineethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO4S · xH2O
CAS Number:
Molecular Weight:
195.24 (anhydrous basis)
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue (900 °C)

≤0.05%

pH

2.5-4.0 (25 °C, 0.5 M in H2O)

useful pH range

5.5-6.7

pKa 

6.1

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.025
λ: 280 nm Amax: 0.020

SMILES string

O.OS(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.H2O/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);1H2

InChI key

MIIIXQJBDGSIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

MES is one of the "Good" buffers developed for biological applications. MES is not recommended at pH 7.4; other buffers should be considered. All qualities are tested for trace ions and all can be used for biological and biochemistry applications. This product is used in biochemical and biological techniques where highly purified products are needed.

Preparation Note

A buffer using MES free acid can be prepared by titrating the free acid with sodium hydroxide to the desired pH (pKa=±1). Alternatively, volumes of equimolar MES free acid and sodium MES can be mixed to attain the desired pH. Standard mixing tables using stock solutions to prepare a buffer of a given pH have been published.

Other Notes

Solubility/Stability: MES is soluble in water, giving a clear colourless solution at concentrations of 0.5M or higher. The pH of a solution should be between 2.5 and 5, depending on the concentration. Solutions are stable at 2-8°C for months.
Sterilization: Sterilization should be by filteration through 0.2μM filters. Autoclaving is not recommended by any sulfonic acid buffers. If buffers must be nuclease-free, it is best to treat the water, then add the buffer solids after autoclaving. When MES solutions are autoclaved, they turn yellow (although pH does not change measurably. The identity of the yellow breakdown product is unknown.
Biological buffer; useful pH-range 5.8-6.5citation; Mobile phase component in brain tubulin isolation by amino-activated gel chromatographycitation; Buffer in plant tissue culturecitationcitation.
Easily compare specifications for MES hydrate products with the MES hydrate specification table.
Biological buffer; useful pH-range 5.8-6.5; Mobile phase component in brain tubulin isolation by amino-activated gel chromatography; Buffer in plant tissue culture.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lacey et al.
Journal of chromatography, 525(1), 71-84 (1990-01-26)
In the present study, we report the isolation of the acidic structural protein tubulin using a number of amino-activated gels. Crude 100,000 g supernatant derived from sheep brain was applied to gels activated with either aminohexyl, aminoethyl, argininyl, diethylaminoethyl, lysinyl
M De Block
Plant physiology, 93(3), 1110-1116 (1990-07-01)
Tissue culture conditions and transformation have been established for both aspen and poplar. The use of previously described culture conditions resulted in shoot tip necrosis in the shoot cultures and necrosis of stem and leaf explants. Shoot tip necrosis could
D.E. Parfitt
Sci. Hortic., 36, 157-157 (1988)
Dawson, R.M.C. et al.
Data for Biochemical Research, 410- 410 (1987)
Good NE et al.
Biochemistry, 5, 467-477 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service