Skip to Content
Merck
All Photos(1)

Key Documents

55865

Sigma-Aldrich

Digoxigenin NHS-ester

≥80% (HPLC)

Synonym(s):

ε-(Digoxigenin-3-0-acetamido)caproic acid N-hydroxysuccinimide ester, DIG NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H50N2O10
CAS Number:
Molecular Weight:
658.78
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥80% (HPLC)

form

solid

shipped in

wet ice

storage temp.

−20°C

SMILES string

[H][C@]12CC[C@]3([H])[C@]([H])(C[C@@H](O)[C@]4(C)[C@H](CC[C@]34O)C5=CC(=O)OC5)[C@@]1(C)CC[C@@H](C2)OCC(=O)NCCCCCC(=O)ON6C(=O)CCC6=O

InChI

1S/C35H50N2O10/c1-33-13-11-23(45-20-28(39)36-15-5-3-4-6-31(42)47-37-29(40)9-10-30(37)41)17-22(33)7-8-25-26(33)18-27(38)34(2)24(12-14-35(25,34)44)21-16-32(43)46-19-21/h16,22-27,38,44H,3-15,17-20H2,1-2H3,(H,36,39)/t22-,23+,24-,25-,26+,27-,33+,34+,35+/m1/s1

InChI key

KHNDABJZSPPYLE-FUGFVFQCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Digoxigenin NHS-ester has been used in the preparation of streptavidin-digoxigenin conjugate and digoxin-recombinant Ara h1 (peanut component) conjugate for enzyme-linked immunosorbent assay (ELISA) analysis.

Biochem/physiol Actions

Digoxigenin NHS-ester is an activated ester that reacts with amino groups under mild conditions, attaching the digoxigenin moiety to proteins or 5′-amino substituted oligonucleotides Digoxigenin labeling is used to immuno-tag molecules for detection using anti-digoxigenin antibodies with high affinities and specificity.
Digoxigenin, an aglycon, is a pharmacologically active plant toxin. It is less potent than digoxin. It is used in lower doses to treat heart conditions. Digoxigenin is usually conjugated to another payload which reduces its remaining activity or accessibility to tissues. Since digoxigenin is permanently bound and entirely covered by the capture modules, it cannot be used as an active entity.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotype-specific agglutination-PCR (ISAP): A sensitive and multiplex method for measuring allergen-specific IgE.
Cheng-Ting Tsai et al.
The Journal of allergy and clinical immunology, 141(5), 1901-1904 (2017-12-19)
R Antolovic et al.
European journal of biochemistry, 227(1-2), 61-67 (1995-01-15)
Na+/K(+)-ATPase from pig kidney is inactivated by protein-reactive N-hydroxysuccinimidyl derivatives of digoxigenin. Like digoxigenin, its protein-reactive derivatives N-hydroxysuccinimidyl digoxigenin-3-methylcarbonyl-epsilon-aminocaproate (HDMA), 3-amino-3-deoxydigoxigenin hemisuccinimide succinimidyl ester (ADHS), 3-iodoacetylamino-3-deoxydigoxigenin (IAD) and digoxigenin-3-O-succinyl-[2-(N-maleimido)]ethylamide (DSME) inhibited the sodium pump in the presence of Na+, Mg2+
Silke Metz et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(20), 8194-8199 (2011-05-04)
Bispecific antibodies that bind cell-surface targets as well as digoxigenin (Dig) were generated for targeted payload delivery. Targeting moieties are IgGs that bind the tumor antigens Her2, IGF1R, CD22, or LeY. A Dig-binding single-chain Fv was attached in disulfide-stabilized form
R Antolovic et al.
FEBS letters, 368(1), 169-172 (1995-07-10)
The digoxigenin derivative N-hydroxysuccinimidyl digoxigenin-3-O-methylcarbonyl-epsilon-aminocaproate (HDMA) has been shown to covalently label the ouabain binding site of the Na,K-ATPase epsilon subunit [Antolovic et al. (1995) Eur. J. Biochem. 227, 61-67]. In the present study we observed both, labeling and inactivation
R Antolovic et al.
The Journal of biological chemistry, 273(26), 16259-16264 (1998-06-20)
Searching for a binding protein in blood, which may be involved in the specific transport of cardiac glycosides to their receptor sites on the sodium pump, we isolated a cardiac glycoside-binding protein (CGBG) of 26 kDa from the globulin fraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service