Skip to Content
Merck
All Photos(3)

Key Documents

482R-1

Sigma-Aldrich

p63 (EP174) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP174

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (482R-14)
vial of 0.1 mL concentrate, Research Use Only (482R-14-RUO)
vial of 0.5 mL concentrate (482R-15)
vial of 1.0 mL concentrate (482R-16)
vial of 1.0 mL concentrate, Research Use Only (482R-16-RUO)
vial of 1.0 mL pre-dilute ready-to-use, Research Use Only (482R-17-RUO)
vial of 1.0 mL pre-dilute, ready to use (482R-17)
vial of 25.0 mL pre-dilute ready-to-use (482R-10)
vial of 25.0 mL pre-dilute ready-to-use, Research Use Only (482R-10-RUO)
vial of 7.0 mL pre-dilute, ready to use (482R-18)
vial of 7.0 mL pre-dilute, ready to use, Research Use Only (482R-18-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable (1:100-1:200 (concentrated))

isotype

IgG

control

lung (squamous cell carcinoma), prostate

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

p63 is a transcription factor that regulates many cell activities, including cell proliferation, maintenance, differentiation, adhesion, and apoptosis.1 It has been found to be a sensitive and specific marker of myoepithelial and basal cell populations in prostate, breast, skin, and salivary gland. Additionally, p63 has also demonstrated immunohistochemical utility in identifying benign and malignant breast lesions.2 In prostatic carcinomas, it can also be used to recognize benign glands adjacent to malignant glands.3 p63 can also be used to differentiate adenocarcinoma of the lungs, where it is usually negative, from squamous cell carcinoma, which can be useful for advanced stages of the carcinoma where surgery is no longer an option.4

Quality

United States - IVD
Canada - RUO
European Union - IVD
Japan - RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong Jiang et al.
American journal of clinical pathology, 123(2), 231-236 (2005-04-22)
We assessed the usefulness of immunohistochemical analysis with a 3-antibody cocktail (alpha-methylacyl coenzyme A racemase [AMACR, or P504S], 34betaE12, p63) and a double-chromogen reaction for detection of limited prostate cancer in 138 needle biopsy specimens, including 82 with small foci
D Stefanou et al.
Histology and histopathology, 19(2), 465-471 (2004-03-17)
The p63 gene encodes six protein isoforms. The transactivating isoforms have similar actions with p53, while the N-isoforms inhibit transcription activation by p53 and transactivating isoforms. p63 is expressed in stratified epithelia and in basal cells of the prostate and
Ricardo Camilo et al.
Human pathology, 37(5), 542-546 (2006-05-02)
In this study, we sought to validate the importance of p63, CK5/CK6, CK7, and surfactant-A (SP-A) to classify 42 non-small cell lung cancers in autopsy and surgical resection specimens and to study the usefulness of these markers in distinguishing between
A Yang et al.
Molecular cell, 2(3), 305-316 (1998-10-17)
We describe the cloning of p63, a gene at chromosome 3q27-29 that bears strong homology to the tumor suppressor p53 and to the related gene, p73. p63 was detected in a variety of human and mouse tissues, including proliferating basal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service