Skip to Content
Merck
All Photos(1)

Key Documents

47729

Sigma-Aldrich

N-Formyl-Met-Leu-Phe

≥99.0% (TLC), BioXtra

Synonym(s):

Chemotactic peptide, N-Formyl-L-methionyl-L-leucyl-L-phenylalanine, fMLF, fMLP (misnomer but widely used)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31N3O5S
CAS Number:
Molecular Weight:
437.55
Beilstein:
2315783
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)

product line

BioXtra

Quality Level

Assay

≥99.0% (TLC)

form

powder

color

white

solubility

DMF: 50 mg/mL, clear, colorless

application(s)

cell analysis

storage temp.

2-8°C

SMILES string

CSCC[C@H](NC=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C21H31N3O5S/c1-14(2)11-17(23-19(26)16(22-13-25)9-10-30-3)20(27)24-18(21(28)29)12-15-7-5-4-6-8-15/h4-8,13-14,16-18H,9-12H2,1-3H3,(H,22,25)(H,23,26)(H,24,27)(H,28,29)/t16-,17-,18-/m0/s1

InChI key

PRQROPMIIGLWRP-BZSNNMDCSA-N

Gene Information

human ... FPR1(2357)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

NFor-Met-Leu-Phe

Application

1. 4th and 5th applications do not have access
2. 3rd application is not relevant

Biochem/physiol Actions

N-Formyl-Met-Leu-Phe is a potent chemotactic peptide and formyl peptide receptor (FPR) agonist that induces a metabolic burst in macrophages accompanied by an increase in respiratory rate, secretion of lysosomal enzymes, and production of superoxide anion.
Potent chemotactic peptide. Induces a metabolic burst in macrophages accompanied by an increase in respiratory rate, secretion of lysosomal enzymes, and production of superoxide anion.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid L Voskamp et al.
The journal of allergy and clinical immunology. In practice, 3(2), 192-199 (2015-02-03)
Allergic bronchopulmonary aspergillosis (ABPA) often presents with persistently uncontrolled asthma despite the use of corticosteroids and antifungal therapy. Omalizumab is a humanized anti-IgE monoclonal antibody currently used to treat severe asthma. The aim was to assess the clinical and immunologic
C Dahlgren et al.
Blood, 95(5), 1810-1818 (2000-02-26)
A D-methionine-containing peptide, Trp-Lys-Tyr-Met-Val-D-Met-NH(2) (WKYMVm), featuring a unique receptor specificity was investigated with respect to its ability to activate neutrophil effector functions. The peptide was found to be more potent than the N-formylated peptide N-formyl-Met-Leu-Phe (fMLF) at inducing neutrophil chemotaxis
J James B Edelman et al.
Shock (Augusta, Ga.), 39(2), 149-154 (2013-01-18)
Persistent alteration to host polymorphonuclear cell (PMN) physiology has been demonstrated after cardiac surgery performed with cardiopulmonary bypass (CPB). However, to date, PMN physiology and function beyond the first 24 h have not been investigated after cardiac surgery performed without
Karama Makni-Maalej et al.
Journal of immunology (Baltimore, Md. : 1950), 189(9), 4657-4665 (2012-09-25)
Superoxide anion production by the neutrophil NADPH oxidase plays a key role in host defense; however, excessive superoxide production is believed to participate to inflammatory reactions. Neutrophils express several TLR that recognize a variety of microbial motifs or agonists. The
Lunhua Liu et al.
Molecular biology of the cell, 23(24), 4751-4765 (2012-10-26)
Integrin activation is required to facilitate multiple adhesion-dependent functions of neutrophils, such as chemotaxis, which is critical for inflammatory responses to injury and pathogens. However, little is known about the mechanisms that mediate integrin activation in neutrophils. We show that

Questions

1–3 of 3 Questions  
  1. How long can this product be stored at -20 when reconstituted in DMSO?

    1 answer
    1. The stability of this product in solution is not determined. However, similar product F3506 states that solutions of N-Formyl-Met-Leu-Phe can be stored, in aliquots, at -20°C for at least six months.

      Helpful?

  2. What is the solubility of the product  47729, N-Formyl-Met-Leu-Phe?

    1 answer
    1. Product is soluble in water (partly), methanol, DMSO (4 mg/ml), ethanol (2 mg/ml), and DMF (Dimethylformamide) (50 mg/ml).

      Helpful?

  3. Can this product be dissolved in DMSO?

    1 answer
    1. Yes, this product can be solubilized in DMSO. The exact concentration at which it may be solubilized is not determined. However, several sources state that it is soluble in DMSO at 30 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service