Skip to Content
Merck
All Photos(1)

Key Documents

45348

Sigma-Aldrich

Epoxy embedding medium, accelerator

≥95% (NT)

Synonym(s):

2,4,6-Tris(dimethylaminomethyl)phenol, Epon DMP 30 substitute, DMP-30

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NCH2]3C6H2OH
CAS Number:
Molecular Weight:
265.39
Beilstein:
795751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.47

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 21 °C)

Assay

≥95% (NT)

form

solid

color

colorless to light yellow

refractive index

n20/D 1.516 (lit.)
n20/D 1.517

bp

130-135 °C/1 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

application(s)

hematology
histology

storage temp.

2-8°C

SMILES string

CN(C)Cc1cc(CN(C)C)c(O)c(CN(C)C)c1

InChI

1S/C15H27N3O/c1-16(2)9-12-7-13(10-17(3)4)15(19)14(8-12)11-18(5)6/h7-8,19H,9-11H2,1-6H3

InChI key

AHDSRXYHVZECER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Epoxy embedding medium, accelerator is used according to the protocol provided for Epoxy Embedding Medium kit, Prod. No. 45359 available on-line. The embedding media is used for electron microscopy.

Legal Information

Epon is a trademark of Hexion, Inc.

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Yamaguchi et al.
Journal of electron microscopy, 58(4), 261-266 (2009-03-18)
A smart and efficient method for freeze substitution and serial sectioning of yeast cells is described. Yeast cells were placed in a single layer between two copper disks, rapidly frozen, freeze substituted and embedded in an epoxy resin. The cell
C K E Bleck et al.
Journal of microscopy, 237(1), 23-38 (2010-01-09)
Bacteria are generally difficult specimens to prepare for conventional resin section electron microscopy and mycobacteria, with their thick and complex cell envelope layers being especially prone to artefacts. Here we made a systematic comparison of different methods for preparing Mycobacterium
S Bratosin-Guttman
Journal of structural biology, 108(2), 162-167 (1992-03-01)
Electron microscopic studies of DNA are hampered by difficulties encountered with the spreading of DNA under protein-free conditions. The established and currently popular technique of spreading DNA on carbon membranes treated by glow discharge in an atmosphere of pentylamine is
A Schaper et al.
FEBS letters, 355(1), 91-95 (1994-11-21)
We present recent advances in DNA specimen preparation technique for scanning force microscopy (SFM) based on spreading on mica in the presence of cationic and non-ionic detergents. Reproducible DNA imaging in air and in n-propanol has been achieved in the
S H Brorson
Micron (Oxford, England : 1993), 29(2-3), 89-95 (1998-07-31)
The purpose of this study was to examine how antigen retrieval affected the yield of immunogold labeling on epoxy sections based on embedding with different amounts of accelerator. The concentration of accelerator DMP-30 (tri(dimethyl amino methyl) phenol) was varied in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service