Skip to Content
Merck
All Photos(1)

Key Documents

43060

Sigma-Aldrich

N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate

BioReagent, suitable for fluorescence, ≥96.0% (TLC)

Synonym(s):

1,6-Diphenyl-1,3,5-hexatriene-4′-trimethylammonium tosylate, 1-(4-Trimethylammoniophenyl)-6-phenyl-1,3,5-hexatriene p-toluenesulfonate, 4′-(Trimethylammonio)diphenylhexatriene p-toluenesulfonate, TMA-DPH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H24N · C7H7O3S
CAS Number:
Molecular Weight:
461.62
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥96.0% (TLC)

form

powder

mp

237 °C (dec.) (lit.)

solubility

DMF: soluble
DMSO: soluble
methanol: soluble

fluorescence

λex 355 nm; λem ~430 nm in methanol
λex 355 nm; λem ~430 nm in methanol

suitability

suitable for fluorescence

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.C[N+](C)(C)c2ccc(\C=C\C=C\C=C\c3ccccc3)cc2

InChI

1S/C21H24N.C7H8O3S/c1-22(2,3)21-17-15-20(16-18-21)14-8-5-4-7-11-19-12-9-6-10-13-19;1-6-2-4-7(5-3-6)11(8,9)10/h4-18H,1-3H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1/b5-4+,11-7+,14-8+;

InChI key

ZKARERKEBVSZCX-VMDDUYISSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N,N,N-Trimethyl-4-(6-phenyl-1,3,5-hexatrien-1-yl)phenylammonium p-toluenesulfonate(TMA-DPH) is used as a fluorescence anisotropy probe in lipid membrane structural studies.

Other Notes

Probe for membrane fluidity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Seydlová et al.
Folia microbiologica, 53(4), 303-307 (2008-09-02)
Processes occurring in the cytoplasmic membrane of the surfactin producer Bacillus subtilis were examined during a 3-d cultivation. The fatty acid composition was found to be almost stable within this interval, except for the early stationary phase when the nonbranched
P Gantet et al.
Plant physiology, 94(2), 729-737 (1990-10-01)
The blue emission anisotropy, r, of two lipophilic probes, diphenylhexatriene (DPH) and its trimethyl-ammonium derivative (TMA-DPH), has been measured in foliar Lupinus albus L. protoplasts for the first time by flow cytometry. Distinctive values have been obtained for protoplasts of
Jacek Grebowski et al.
Biochimica et biophysica acta, 1828(2), 241-248 (2012-09-20)
The influence of fullerenol on the activities of human erythrocyte membrane ATPases and the fluidity of the plasma membrane as well as the possibility of fullerenol incorporation into the plasma membrane were investigated. Fullerenol at concentrations up to 150 μg/mL
Maria Stasiuk et al.
FEBS letters, 582(25-26), 3607-3613 (2008-10-07)
The effects induced by natural phenolic and resorcinolic lipids on membrane permeability were investigated. All of the compounds tested perturbed the phospholipid bilayer and stabilized erythrocytes against hypoosmotically induced hemolysis. Dipalmitoylphosphatidylcholine liposomes with two preincorporated fluorescent dyes (1-(4-trimethylammoniumphenyl)-6-phenyl-1,3,5-hexatrien p-toluenesulfonate (TMA-DPH)
Eugenia Kovács et al.
Bioelectrochemistry (Amsterdam, Netherlands), 87, 230-235 (2012-04-24)
The interaction of positively-charged antibiotic gentamicin with cell membranes was studied to determine if any changes in membrane organization were induced by the drug. Opossum kidney epithelia (OK) cells were used as models of eukaryotic cells. Two methods were used:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service