Skip to Content
Merck
All Photos(1)

Key Documents

41785

Sigma-Aldrich

Dimidium bromide

BioReagent, suitable for fluorescence, ~95% (AT)

Synonym(s):

3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide, Trypadine, phenanthridinium 1553

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H18BrN3
CAS Number:
Molecular Weight:
380.28
Beilstein:
3809779
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32
Assay:
~95% (AT)
form:
solid

product line

BioReagent

Quality Level

Assay

~95% (AT)

form

solid

mp

243-248 °C (lit.)

fluorescence

λex 306 nm; λem 612 nm in 50 mM Tris pH 8.0 (DNA)
λex 480 (pH 7.6, low ionic strength)

suitability

suitable for fluorescence

SMILES string

[Br-].C[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C20H17N3.BrH/c1-23-19-12-15(22)8-10-17(19)16-9-7-14(21)11-18(16)20(23)13-5-3-2-4-6-13;/h2-12,22H,21H2,1H3;1H

InChI key

MQOKYEROIFEEBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimidium Bromide is a trypanocidal drug that is known to affect nucleic acid synthesis in many organisms e.g. tissue culture cells.

Application

Dimidium Bromide is used as a binding agent to natural DNA based on base-pair specificity. It is used as a fluorescence agent for nucleic acid experiments.
Intercalating probe for nucleic acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Dougherty
The International journal of biochemistry, 14(6), 493-504 (1982-01-01)
1. The absorption spectrum of three phenanthridine drugs (ethidium, dimidium and prothidium bromide) bound to natural DNAs of differing G-C content were obtained using a novel mixing scheme and analysed according to the excluded site binding model. 2. Ethidium bromide
Nuclear transplantation studies of the action of dimidium bromide on amoebae.
S E Hawkins et al.
Nature, 222(5188), 86-87 (1969-04-05)
[Chemoprophylaxis of bovine trypanosomiasis T. congolense and T. vivax by phenathridinium S. 1553].
J DEOM
Annales de la Societe belge de medecine tropicale (1920), 30(1), 25-29 (1950-03-01)
T N Beliaeva et al.
Tsitologiia, 31(11), 1363-1368 (1989-11-01)
A natural DNA-intercalator plant benzo-c-phenanthridine alkaloid sanguinarine is more toxic for mouse transformed fibroblast L-cells in culture than synthetic DNA-intercalator ethidium bromide (EtB) and alkaloid berberine. Dimidium bromide is also an inhibitor of the L-cell growth. In assay conditions, growth
Dimidium bromide or phenanthridinium 1553; a note on the present position.
J CARMICHAEL
The Veterinary record, 62(17), 257-257 (1950-04-29)

Related Content

Nancy-520 for DNA Detection and Quantitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service