Skip to Content
Merck
All Photos(1)

Key Documents

3871

Sigma-Aldrich

Naphthol AS-BI Phosphoric Acid Solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

12.5 mg/mL

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

SMILES string

COc1ccccc1NC(=O)c2cc3cc(Br)ccc3cc2OP(O)(O)=O

InChI

1S/C18H15BrNO6P/c1-25-16-5-3-2-4-15(16)20-18(21)14-9-12-8-13(19)7-6-11(12)10-17(14)26-27(22,23)24/h2-10H,1H3,(H,20,21)(H2,22,23,24)

InChI key

HUXIAXQSTATULQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For use in the Sigma Acid Phosphatase (TRAP) Procedure, Kit 387A

Components

Napthol AS-BI phosphoric acid, 12.5 mg/mL.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronika Kozlova et al.
PloS one, 15(3), e0229170-e0229170 (2020-03-27)
Surface protein CD20 serves as the critical target of immunotherapy in various B-cell malignancies for decades, however its biological function and regulation remain largely elusive. Better understanding of CD20 function may help to design improved rational therapies to prevent development
Yi-Ju Pan et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 67(10), 847-861 (2018-08-16)
To investigate the amelioration effects of quetiapine on rheumatoid arthritis with RAW 264.7 macrophage and collagen-induced arthritis (CIA) DBA/1J mouse model. RAW 264.7 macrophage and DBA/1J mice. Lipopolysaccharide and collagen. RAW 264.7 macrophages stimulated by lipopolysaccharide (LPS) followed by quetiapine
Daniel P Duarte et al.
Nature communications, 11(1), 2319-2319 (2020-05-10)
Bruton's tyrosine kinase (Btk) is critical for B-cell maturation and activation. Btk loss-of-function mutations cause human X-linked agammaglobulinemia (XLA). In contrast, Btk signaling sustains growth of several B-cell neoplasms which may be treated with tyrosine kinase inhibitors (TKIs). Here, we
Qi Xiong et al.
Proteome science, 13, 16-16 (2015-05-15)
Cultivation of osteoclasts is a basic tool for investigating osteolytic bone diseases. Fetal bovine serum (FBS) is the standard supplement used for in vitro cell culture medium. Typically, the serum volume fraction used for osteoclast cultivation is 10%. In this
Qi Xiong et al.
BioMed research international, 2015, 596789-596789 (2015-06-30)
Estrogen plays an important role in inhibiting osteoclast differentiation and protecting against bone loss from osteoporosis, especially in postmenopausal women. However, the precise mechanisms underlying the effect of estrogen on osteoclasts are not well known. In the present study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service