Skip to Content
Merck
All Photos(1)

Key Documents

367R-1

Sigma-Aldrich

IgG4 (EP138) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP138, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (367R-17)
bottle of 7.0 mL predilute (367R-18)
vial of 0.1 mL concentrate (367R-14)
vial of 0.5 mL concentrate (367R-15)
vial of 1.0 mL concentrate (367R-16)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

IgG4-related sclerosing disease has been recognized as a systemic disease entity characterized by an elevated serum IgG4 level, sclerosing fibrosis, and diffuse lymphoplasmacytic infiltration with the presence of many IgG4-positive plasma cells. Clinical manifestations are apparent in the pancreas, bile duct, gall bladder, lacrimal gland, salivary gland, retroperitoneum, kidney, lung, breast, thyroid, and prostate. Immunohistochemical analyses in the case of IgG4-related sclerosing disease not only exhibit significantly more than normal IgG4-positive plasma cells in affected tissues but also significantly higher IgG4/IgG ratios (typically > 30%).1-8

Quality


IVD

IVD

IVD

RUO

Linkage

IgG4 Positive Control Slides, Product No. 367S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuharu Sato et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(4), 589-599 (2009-03-10)
IgG4-related disease sometimes involves regional and/or systemic lymph nodes, and often clinically and/or histologically mimics multicentric Castleman's disease or malignant lymphoma. In this study, we examined clinical and pathologic findings of nine patients with systemic IgG4-related lymphadenopathy. None of these
Wah Cheuk et al.
The American journal of surgical pathology, 33(7), 1058-1064 (2009-04-23)
Immunoglobulin G (IgG)4-related sclerosing disease is a recently described syndrome characterized by mass-forming lesions in various organs due to dense lymphoplasmacytic infiltrates and stromal sclerosis, elevated serum IgG4 titer, increased tissue IgG4 plasma cells, and favorable clinical outcome. We describe
Terumi Kamisawa et al.
World journal of gastroenterology, 15(19), 2357-2360 (2009-05-20)
To clarify the characteristic features of biliary lesions in patients with autoimmune pancreatitis (AIP) and compare them with those of primary sclerosing cholangitis (PSC). The clinicopathological characteristics of 34 patients with sclerosing cholangitis (SC) associated with AIP were compared with
Yaqiong Li et al.
Pathology international, 59(9), 636-641 (2009-08-29)
IgG4-related sclerosing disease has been recently recognized as a systemic disease entity characterized by an elevated serum IgG4 level, sclerosing fibrosis and diffuse lymphoplasmacytic infiltration by many IgG4-positive plasma cells. Similar histopathological features have often been noted in the fibrous
Vikram Deshpande et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(10), 1287-1295 (2009-07-28)
IgG4-associated cholangitis is a steroid-responsive hepatobiliary inflammatory condition associated with autoimmune pancreatitis that clinically and radiologically mimics primary sclerosing cholangitis. In this study, we conducted a morphological and immunohistochemical analysis of liver material obtained from individuals with IgG4-associated cholangitis, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service