Skip to Content
Merck
All Photos(1)

Key Documents

33211

Sigma-Aldrich

Cadaverine

purum, ≥97.0% (GC)

Synonym(s):

1,5-Diaminopentane, 1,5-Pentanediamine, Pentamethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)5NH2
CAS Number:
Molecular Weight:
102.18
Beilstein:
1697256
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.458 (lit.)
n20/D 1.458

bp

178-180 °C (lit.)

mp

14-16 °C (lit.)

solubility

1 M HCl: soluble 0.5 g/10 mL, clear, colorless

density

0.869 g/mL at 20 °C
0.873 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

NCCCCCN

InChI

1S/C5H14N2/c6-4-2-1-3-5-7/h1-7H2

InChI key

VHRGRCVQAFMJIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cadaverine is a biogenic amine.[1] Mechanism of cadaverine fermentation by Corynebacterium glutamicum was reported.[2]
Cadaverine, a diamine, has been reported as a cadaveric volatile organic compound. Its effect on the electroantennographic (EAG) and behavioral responses of silphine species, Thanatophilus sinuatus Fabricius has been investigated.[3]

Application

Cadaverine has been used as diamine in the synthesis of diamine-inorganic anion [Cl-, SO42-, HPO42-, P2O74- and Fe(CN)64-] complexes.[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophysiological and behavioral responses of Thanatophilus sinuatus Fabricius (Coleoptera: Silphidae) to selected cadaveric volatile organic compounds.
Dekeirsschieter J, et al.
Journal of Forensic Sciences, 58(4), 917-923 (2013)
Biljana R Cvetković et al.
Food chemistry, 168, 471-477 (2014-08-31)
White cabbage heads cultivar "Futoški" and hybrid "Bravo" were investigated during fermentation process, for 50days, at different temperature regimes (16-18; 18-20; 20-22°C) and salt concentrations 1, 1.5 and 2%. The quantity of biogenic amines (tryptamine, phenylethylamine, putrescine, cadaverine, histamine, serotonine
Takashi Mimitsuka et al.
Bioscience, biotechnology, and biochemistry, 71(9), 2130-2135 (2007-09-27)
Cadaverine, the expected raw material of polyamides, is produced by decarboxylation of L-lysine. If we could produce cadaverine from the cheapest sugar, and as a renewable resource, it would be an effective solution against global warming, but there has been
S Kubota et al.
Cancer research, 43(5), 2363-2367 (1983-05-01)
We developed a new simple enzymatic assay method for measuring urinary polyamines (total amount of putrescine, spermidine, and cadaverine), using an acylpolyamine amidohydrolase and a putrescine oxidase. First conjugated polyamines (putrescine, spermidine, and cadaverine) in urine were hydrolyzed by incubation
S Goldberg et al.
Journal of dental research, 73(6), 1168-1172 (1994-06-01)
Whereas previous studies have shown correlations between volatile sulphur compounds (VSC) and bad breath levels, it is probable that other compounds found in the oral cavity may contribute to oral malodor. In the present investigation, the possibility that diamines (cadaverine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service